Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05020554.hg.1 |
TC05001502.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05020554.hg.1 |
ENST00000296641.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05020554.hg.1 |
ENST00000504899.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05020554.hg.1 |
NM_004101.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011194.hg.1 |
ENST00000296641.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011194.hg.1 |
ENST00000296641.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011194.hg.1 |
ENST00000296641.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011194.hg.1 |
ENST00000296641.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011194.hg.1 |
ENST00000296641.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011194.hg.1 |
ENST00000296641.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011194.hg.1 |
ENST00000296641.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011194.hg.1 |
ENST00000296641.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011194.hg.1 |
ENST00000504899.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011194.hg.1 |
ENST00000504899.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011194.hg.1 |
ENST00000504899.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011194.hg.1 |
ENST00000504899.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011194.hg.1 |
ENST00000504899.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011194.hg.1 |
NM_001256566.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011194.hg.1 |
NM_001256566.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011194.hg.1 |
NM_001256566.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011194.hg.1 |
NM_001256566.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011194.hg.1 |
NM_001256566.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011194.hg.1 |
NM_001256566.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011194.hg.1 |
NM_004101.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011194.hg.1 |
NM_004101.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011194.hg.1 |
NM_004101.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011194.hg.1 |
NM_004101.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011194.hg.1 |
NM_004101.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011194.hg.1 |
NM_004101.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011194.hg.1 |
NM_004101.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011194.hg.1 |
NM_004101.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05020551.hg.1 |
TC05001502.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05020551.hg.1 |
ENST00000296641.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05020551.hg.1 |
ENST00000504899.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05020551.hg.1 |
NM_004101.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05020552.hg.1 |
TC05001502.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05020552.hg.1 |
ENST00000296641.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05020552.hg.1 |
NM_004101.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05020553.hg.1 |
TC05001502.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05020553.hg.1 |
ENST00000296641.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05020553.hg.1 |
ENST00000504899.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05020553.hg.1 |
NM_004101.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500177647.hg.1 |
ENST00000296641.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500177647.hg.1 |
NM_004101.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500177647.hg.1 |
ENST00000296641.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500177647.hg.1 |
NM_004101.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500177647.hg.1 |
TC0500011194.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500177648.hg.1 |
ENST00000296641.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500177648.hg.1 |
ENST00000504899.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500177648.hg.1 |
NM_001256566.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500177648.hg.1 |
NM_004101.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500177648.hg.1 |
ENST00000296641.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500177648.hg.1 |
ENST00000504899.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500177648.hg.1 |
NM_001256566.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500177648.hg.1 |
NM_004101.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500177648.hg.1 |
TC0500011194.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500177646.hg.1 |
ENST00000296641.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500177646.hg.1 |
NM_004101.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500177646.hg.1 |
ENST00000296641.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500177646.hg.1 |
NM_004101.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500177646.hg.1 |
TC0500011194.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500177649.hg.1 |
ENST00000296641.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500177649.hg.1 |
ENST00000504899.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500177649.hg.1 |
NM_001256566.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500177649.hg.1 |
NM_004101.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500177649.hg.1 |
ENST00000296641.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500177649.hg.1 |
ENST00000504899.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500177649.hg.1 |
NM_001256566.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500177649.hg.1 |
NM_004101.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500177649.hg.1 |
TC0500011194.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500177644.hg.1 |
ENST00000296641.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500177644.hg.1 |
ENST00000504899.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500177644.hg.1 |
NM_001256566.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500177644.hg.1 |
NM_004101.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500177644.hg.1 |
ENST00000296641.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500177644.hg.1 |
ENST00000504899.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500177644.hg.1 |
NM_001256566.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500177644.hg.1 |
NM_004101.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500177644.hg.1 |
TC0500011194.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500011194.hg.1 |
ENST00000296641.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011194.hg.1 |
ENST00000296641.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011194.hg.1 |
ENST00000296641.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011194.hg.1 |
ENST00000296641.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011194.hg.1 |
ENST00000296641.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011194.hg.1 |
ENST00000296641.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011194.hg.1 |
ENST00000296641.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011194.hg.1 |
ENST00000296641.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011194.hg.1 |
ENST00000504899.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011194.hg.1 |
ENST00000504899.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011194.hg.1 |
ENST00000504899.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011194.hg.1 |
ENST00000504899.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011194.hg.1 |
ENST00000504899.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011194.hg.1 |
NM_001256566.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011194.hg.1 |
NM_001256566.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011194.hg.1 |
NM_001256566.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011194.hg.1 |
NM_001256566.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011194.hg.1 |
NM_001256566.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011194.hg.1 |
NM_001256566.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011194.hg.1 |
NM_004101.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011194.hg.1 |
NM_004101.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011194.hg.1 |
NM_004101.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011194.hg.1 |
NM_004101.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011194.hg.1 |
NM_004101.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011194.hg.1 |
NM_004101.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011194.hg.1 |
NM_004101.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011194.hg.1 |
NM_004101.3 |
Refseq |