Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02001328.hg.1 |
TC02000119.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02001328.hg.1 |
ENST00000272224.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02001328.hg.1 |
NM_182828.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02001326.hg.1 |
TC02000119.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02001326.hg.1 |
ENST00000272224.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02001326.hg.1 |
NM_182828.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02001327.hg.1 |
TC02000119.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02001327.hg.1 |
ENST00000272224.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02001327.hg.1 |
NM_182828.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006900.hg.1 |
ENST00000272224.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006900.hg.1 |
ENST00000272224.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006900.hg.1 |
ENST00000272224.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006900.hg.1 |
ENST00000272224.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006900.hg.1 |
ENST00000272224.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006900.hg.1 |
ENST00000272224.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006900.hg.1 |
ENST00000565841.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006900.hg.1 |
NM_182828.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006900.hg.1 |
NM_182828.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006900.hg.1 |
NM_182828.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006900.hg.1 |
NM_182828.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200147103.hg.1 |
ENST00000272224.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200147103.hg.1 |
NM_182828.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200147103.hg.1 |
ENST00000272224.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200147103.hg.1 |
NM_182828.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200147103.hg.1 |
TC0200006900.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200147100.hg.1 |
ENST00000272224.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200147100.hg.1 |
NM_182828.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200147100.hg.1 |
ENST00000272224.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200147100.hg.1 |
NM_182828.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200147100.hg.1 |
TC0200006900.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200006900.hg.1 |
ENST00000272224.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006900.hg.1 |
ENST00000272224.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006900.hg.1 |
ENST00000272224.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006900.hg.1 |
ENST00000272224.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006900.hg.1 |
ENST00000272224.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006900.hg.1 |
ENST00000272224.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006900.hg.1 |
ENST00000565841.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006900.hg.1 |
NM_182828.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006900.hg.1 |
NM_182828.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006900.hg.1 |
NM_182828.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006900.hg.1 |
NM_182828.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200147102.hg.1 |
ENST00000272224.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200147102.hg.1 |
NM_182828.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200147102.hg.1 |
ENST00000272224.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200147102.hg.1 |
NM_182828.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200147102.hg.1 |
TC0200006900.hg.1 |
Clariom D TC |