Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCE1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178349.1 | NP_848126.1 | 1 | 926 | 128 | |
GenBank mRNA | BC031811.1 | - | 1 | 926 | 128 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens 1700 Inventoried Both primers and probe map within a single exon |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00513640 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20570795 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20570774 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20570785 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20570776 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00513633 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00513639 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20570780 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15129437 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15129427 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20570783 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20570779 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15129432 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20570773 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15129428 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00513631 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20570778 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20570784 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 101180 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 310531 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3356816 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P368862 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 36734 | - | - |
Agilent Human Genome CGH Microarray 44B | A_14_P134645 | - | - |
Agilent Human Genome CGH Microarray 44A | A_14_P134645 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 62469 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_16_P15297278 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3356816 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3356816 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00513640 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20570778 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20570774 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00513637 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20570776 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20570795 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15129437 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00513639 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20570773 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00513631 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20570780 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20570779 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20570784 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20570796 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15129432 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20570783 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00513633 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20570774 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20570795 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00513633 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20570784 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20570778 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20570796 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00513631 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15129432 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00513640 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20570776 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00513637 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20570773 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15129437 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20570783 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20570779 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00513639 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20570780 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_23_P368862 | - | - |
Agilent Human Genome CGH Microarray 105A (G4412A) | A_14_P134645 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00513639 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00513637 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00513635 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00513642 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00513640 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_14_P134645 | - | - |
Agilent Human Genome CGH Microarray 4x44K | A_14_P134645 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P368862 | - | - |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 103960 | - | - |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 476 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P15297278 | - | - |
Agilent Human 1B Microarray G4111A | A_32_P779544 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00513590 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00513637 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00513593 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00513608 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00513610 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00513622 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00513639 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15129432 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00513603 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15129431 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00513657 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00513600 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00513628 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15129415 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00513576 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00513631 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00513613 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00513595 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00513605 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15129426 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00513582 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00513581 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15129417 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15129437 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00513625 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00513579 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00513578 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00513598 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00513616 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00513611 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00513662 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00513592 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00513629 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15129439 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00513642 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15129414 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00513635 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00513619 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00513660 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P15297278 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | NM_178349_at | - | - |
Affymetrix Human Gene 1.0 ST Array | 7905527 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7905526 | - | - |
Affymetrix Human Gene 1.1 ST Array | 7905525 | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 1560531_PM_at | - | - |
Affymetrix Human Genome U219 Array | 11737945_x_at | - | - |
Affymetrix Human Genome U219 Array | 11737945_PM_x_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | PSR01019047.hg.1 | TC01001240.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01019047.hg.1 | ENST00000360090.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01019047.hg.1 | ENST00000439693.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01019047.hg.1 | NM_178349.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01019049.hg.1 | TC01001240.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01019049.hg.1 | ENST00000360090.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01019049.hg.1 | ENST00000439693.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01019049.hg.1 | NM_178349.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01019046.hg.1 | TC01001240.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01019046.hg.1 | ENST00000360090.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01019046.hg.1 | NM_178349.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01019051.hg.1 | TC01001240.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01019051.hg.1 | ENST00000360090.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01019051.hg.1 | NM_178349.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018297.hg.1 | ENST00000615904.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018297.hg.1 | ENST00000615904.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01019044.hg.1 | TC01001240.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01019044.hg.1 | ENST00000360090.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01019044.hg.1 | NM_178349.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01019050.hg.1 | TC01001240.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01019050.hg.1 | ENST00000360090.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01019050.hg.1 | NM_178349.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018296.hg.1 | ENST00000360090.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018296.hg.1 | ENST00000360090.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018296.hg.1 | ENST00000360090.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018296.hg.1 | ENST00000360090.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018296.hg.1 | ENST00000360090.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018296.hg.1 | NM_178349.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018296.hg.1 | NM_178349.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018296.hg.1 | NM_178349.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018296.hg.1 | NM_178349.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018296.hg.1 | NM_178349.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100239454.hg.1 | ENST00000360090.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100239454.hg.1 | NM_178349.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100239454.hg.1 | ENST00000360090.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100239454.hg.1 | NM_178349.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100239454.hg.1 | TC0100018296.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0100018297.hg.1 | ENST00000615904.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018297.hg.1 | ENST00000615904.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100239455.hg.1 | ENST00000360090.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100239455.hg.1 | NM_178349.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100239455.hg.1 | ENST00000360090.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100239455.hg.1 | NM_178349.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100239455.hg.1 | TC0100018296.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100239453.hg.1 | ENST00000360090.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100239453.hg.1 | NM_178349.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100239453.hg.1 | ENST00000360090.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100239453.hg.1 | NM_178349.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100239453.hg.1 | TC0100018296.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100239456.hg.1 | ENST00000360090.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100239456.hg.1 | NM_178349.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100239456.hg.1 | ENST00000360090.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100239456.hg.1 | NM_178349.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100239456.hg.1 | TC0100018296.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0100018296.hg.1 | ENST00000360090.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018296.hg.1 | ENST00000360090.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018296.hg.1 | ENST00000360090.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018296.hg.1 | ENST00000360090.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018296.hg.1 | ENST00000360090.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018296.hg.1 | NM_178349.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018296.hg.1 | NM_178349.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018296.hg.1 | NM_178349.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018296.hg.1 | NM_178349.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018296.hg.1 | NM_178349.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100239451.hg.1 | ENST00000360090.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100239451.hg.1 | NM_178349.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100239451.hg.1 | ENST00000360090.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100239451.hg.1 | NM_178349.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100239451.hg.1 | TC0100018296.hg.1 | Clariom D TC |
Human Genome U133 Plus 2.0 Array | 1560531_at | - | - |
Human Genome U133 Plus 2.0 Array | 1560531_PM_at | - | - |
Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) | NM_178349_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1560531_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1560531_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1560531_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1560531_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1560531_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1560531_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1560531_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1560531_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1560531_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1560531_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1560531_at6 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A031452 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A031452 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 232514 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_1736655 | NM_178349.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1736655 | BC031811.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1736655 | NM_178349.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1736655 | BC031811.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1736655 | NM_178349.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1736655 | BC031811.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1736655 | NM_178349.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1736655 | BC031811.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1736655 | NM_178349.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1736655 | BC031811.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_30387655-S | NM_178349.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_30387655-S | BC031811.1 | human_transcriptome |
Illumina HumanRef-8 v2 Expression BeadChip | GI_23910-S | - | - |
Illumina human-6 v1.0 expression beadchip | 0003140494 | - | - |
Illumina humanRef-8 v1.0 expression beadchip | 0003140494 | - | - |
Illumina human-6 v2.0 expression beadchip | ILMN_1736655 | NM_178349.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1736655 | BC031811.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1736655 | NM_178349.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1736655 | BC031811.1 | human_transcriptome |
Illumina Sentrix Human-6 v2 Expression BeadChip | 3400554 | - | - |
Illumina HumanRef-6 v2.0 expression beadchip | ILMN_23910 | - | - |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1736655 | NM_178349.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1736655 | BC031811.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1736655 | NM_178349.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1736655 | BC031811.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1736655 | NM_178349.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1736655 | BC031811.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1736655 | NM_178349.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1736655 | BC031811.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1736655 | NM_178349.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1736655 | BC031811.1 | human_transcriptome |