Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YBEY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001314022.1 | NP_001300951.1 | 2 - 3 | 659 | 73 | |
XM_005261156.3 | XP_005261213.2 | 2 - 3 | 611 | 73 | ||
XM_011529634.2 | XP_011527936.2 | 2 - 3 | 611 | 73 | ||
XM_017028394.1 | XP_016883883.1 | 2 - 3 | 611 | 73 | ||
GenBank mRNA | AY040874.1 | - | 2 - 3 | 605 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Homo sapiens Probe spans exons |