Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR08017596.hg.1 |
TC08001364.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08017596.hg.1 |
ENST00000379071.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08017596.hg.1 |
NM_001080526.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010937.hg.1 |
ENST00000379071.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010937.hg.1 |
ENST00000379071.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010937.hg.1 |
ENST00000379071.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010937.hg.1 |
ENST00000379071.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010937.hg.1 |
ENST00000379071.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010937.hg.1 |
ENST00000379071.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010937.hg.1 |
NM_001080526.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010937.hg.1 |
NM_001080526.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010937.hg.1 |
NM_001080526.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010937.hg.1 |
NM_001080526.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08017597.hg.1 |
TC08001364.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08017597.hg.1 |
ENST00000379071.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08017597.hg.1 |
NM_001080526.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010937.hg.1 |
ENST00000379071.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010937.hg.1 |
ENST00000379071.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010937.hg.1 |
ENST00000379071.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010937.hg.1 |
ENST00000379071.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010937.hg.1 |
ENST00000379071.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010937.hg.1 |
ENST00000379071.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010937.hg.1 |
NM_001080526.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010937.hg.1 |
NM_001080526.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010937.hg.1 |
NM_001080526.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010937.hg.1 |
NM_001080526.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800174271.hg.1 |
ENST00000379071.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800174271.hg.1 |
NM_001080526.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800174271.hg.1 |
ENST00000379071.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800174271.hg.1 |
NM_001080526.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800174271.hg.1 |
TC0800010937.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800174272.hg.1 |
ENST00000379071.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800174272.hg.1 |
NM_001080526.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800174272.hg.1 |
ENST00000379071.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800174272.hg.1 |
NM_001080526.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800174272.hg.1 |
TC0800010937.hg.1 |
Clariom D TC |