Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007610.hg.1 |
ENST00000382713.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007610.hg.1 |
ENST00000382713.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007610.hg.1 |
ENST00000382713.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007610.hg.1 |
ENST00000382713.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007610.hg.1 |
ENST00000382713.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007610.hg.1 |
ENST00000382713.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007610.hg.1 |
ENST00000382713.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007610.hg.1 |
ENST00000382713.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007610.hg.1 |
ENST00000382713.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007610.hg.1 |
ENST00000382713.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007610.hg.1 |
ENST00000382713.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007610.hg.1 |
ENST00000382713.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007610.hg.1 |
ENST00000382713.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007610.hg.1 |
ENST00000382713.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007610.hg.1 |
ENST00000577609.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007610.hg.1 |
ENST00000577609.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007610.hg.1 |
ENST00000577609.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007610.hg.1 |
ENST00000577609.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007610.hg.1 |
ENST00000582172.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007610.hg.1 |
ENST00000582172.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007610.hg.1 |
ENST00000582172.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007610.hg.1 |
ENST00000582172.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007610.hg.1 |
ENST00000582992.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007610.hg.1 |
ENST00000582992.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007610.hg.1 |
ENST00000582992.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007610.hg.1 |
ENST00000582992.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007610.hg.1 |
ENST00000582992.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007610.hg.1 |
ENST00000584435.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007610.hg.1 |
ENST00000584435.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007610.hg.1 |
ENST00000584435.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007610.hg.1 |
ENST00000584435.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007610.hg.1 |
ENST00000584435.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007610.hg.1 |
NM_152721.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007610.hg.1 |
NM_152721.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007610.hg.1 |
NM_152721.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007610.hg.1 |
NM_152721.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007610.hg.1 |
NM_152721.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007610.hg.1 |
NM_152721.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007610.hg.1 |
NM_152721.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007610.hg.1 |
NM_152721.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007610.hg.1 |
NM_152721.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007610.hg.1 |
NM_152721.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007610.hg.1 |
NM_152721.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007610.hg.1 |
NM_152721.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007610.hg.1 |
NM_152721.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007610.hg.1 |
NM_152721.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002842.hg.1 |
ENST00000382713.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002842.hg.1 |
NM_152721.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002842.hg.1 |
TC18000241.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1800152167.hg.1 |
ENST00000382713.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800152167.hg.1 |
ENST00000582992.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800152167.hg.1 |
ENST00000584435.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800152167.hg.1 |
NM_152721.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800152167.hg.1 |
TC1800007610.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800152167.hg.1 |
ENST00000382713.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800152167.hg.1 |
ENST00000582992.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800152167.hg.1 |
ENST00000584435.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800152167.hg.1 |
NM_152721.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007610.hg.1 |
ENST00000382713.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007610.hg.1 |
ENST00000382713.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007610.hg.1 |
ENST00000382713.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007610.hg.1 |
ENST00000382713.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007610.hg.1 |
ENST00000382713.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007610.hg.1 |
ENST00000382713.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007610.hg.1 |
ENST00000382713.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007610.hg.1 |
ENST00000382713.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007610.hg.1 |
ENST00000382713.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007610.hg.1 |
ENST00000382713.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007610.hg.1 |
ENST00000382713.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007610.hg.1 |
ENST00000382713.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007610.hg.1 |
ENST00000382713.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007610.hg.1 |
ENST00000382713.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007610.hg.1 |
ENST00000577609.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007610.hg.1 |
ENST00000577609.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007610.hg.1 |
ENST00000577609.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007610.hg.1 |
ENST00000577609.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007610.hg.1 |
ENST00000582172.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007610.hg.1 |
ENST00000582172.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007610.hg.1 |
ENST00000582172.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007610.hg.1 |
ENST00000582172.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007610.hg.1 |
ENST00000582992.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007610.hg.1 |
ENST00000582992.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007610.hg.1 |
ENST00000582992.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007610.hg.1 |
ENST00000582992.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007610.hg.1 |
ENST00000582992.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007610.hg.1 |
ENST00000584435.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007610.hg.1 |
ENST00000584435.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007610.hg.1 |
ENST00000584435.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007610.hg.1 |
ENST00000584435.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007610.hg.1 |
ENST00000584435.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007610.hg.1 |
NM_152721.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007610.hg.1 |
NM_152721.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007610.hg.1 |
NM_152721.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007610.hg.1 |
NM_152721.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007610.hg.1 |
NM_152721.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007610.hg.1 |
NM_152721.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007610.hg.1 |
NM_152721.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007610.hg.1 |
NM_152721.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007610.hg.1 |
NM_152721.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007610.hg.1 |
NM_152721.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007610.hg.1 |
NM_152721.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007610.hg.1 |
NM_152721.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007610.hg.1 |
NM_152721.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007610.hg.1 |
NM_152721.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800152169.hg.1 |
ENST00000382713.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800152169.hg.1 |
ENST00000577609.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800152169.hg.1 |
ENST00000582992.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800152169.hg.1 |
ENST00000584435.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800152169.hg.1 |
NM_152721.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800152169.hg.1 |
TC1800007610.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800152169.hg.1 |
ENST00000382713.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800152169.hg.1 |
ENST00000577609.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800152169.hg.1 |
ENST00000582992.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800152169.hg.1 |
ENST00000584435.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800152169.hg.1 |
NM_152721.5 |
Refseq |