Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007261.hg.1 |
ENST00000356450.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007261.hg.1 |
ENST00000356450.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007261.hg.1 |
ENST00000356450.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007261.hg.1 |
ENST00000356450.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007261.hg.1 |
ENST00000376006.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007261.hg.1 |
ENST00000376006.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007261.hg.1 |
ENST00000376006.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007261.hg.1 |
ENST00000376006.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007261.hg.1 |
NM_001304963.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007261.hg.1 |
NM_001304963.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007261.hg.1 |
NM_001304963.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007261.hg.1 |
NM_001304963.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007261.hg.1 |
NM_153183.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007261.hg.1 |
NM_153183.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007261.hg.1 |
NM_153183.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007261.hg.1 |
NM_153183.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X003893.hg.1 |
TC0X000291.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X003893.hg.1 |
ENST00000356450.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X003893.hg.1 |
ENST00000376006.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X003893.hg.1 |
NM_153183.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X003892.hg.1 |
TC0X000291.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X003892.hg.1 |
ENST00000356450.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X003892.hg.1 |
ENST00000376006.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X003892.hg.1 |
NM_153183.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009694.hg.1 |
ENST00000375992.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009694.hg.1 |
ENST00000375992.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009694.hg.1 |
ENST00000375992.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009694.hg.1 |
NM_018159.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009694.hg.1 |
NM_018159.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009694.hg.1 |
NM_018159.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X003888.hg.1 |
TC0X000291.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X003888.hg.1 |
ENST00000376006.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X003888.hg.1 |
NM_153183.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00149983.hg.1 |
ENST00000356450.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00149983.hg.1 |
ENST00000376006.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00149983.hg.1 |
NM_001304963.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00149983.hg.1 |
NM_153183.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00149983.hg.1 |
ENST00000356450.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00149983.hg.1 |
ENST00000376006.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00149983.hg.1 |
NM_001304963.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00149983.hg.1 |
NM_153183.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00149983.hg.1 |
TC0X00007261.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00149980.hg.1 |
ENST00000356450.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00149980.hg.1 |
NM_001304963.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00149980.hg.1 |
ENST00000356450.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00149980.hg.1 |
NM_001304963.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00149980.hg.1 |
TC0X00007261.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00149978.hg.1 |
ENST00000376006.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00149978.hg.1 |
NM_153183.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00149978.hg.1 |
ENST00000376006.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00149978.hg.1 |
NM_153183.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00149978.hg.1 |
TC0X00007261.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00149982.hg.1 |
ENST00000356450.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00149982.hg.1 |
ENST00000376006.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00149982.hg.1 |
NM_001304963.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00149982.hg.1 |
NM_153183.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00149982.hg.1 |
ENST00000356450.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00149982.hg.1 |
ENST00000376006.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00149982.hg.1 |
NM_001304963.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00149982.hg.1 |
NM_153183.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00149982.hg.1 |
TC0X00007261.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00149985.hg.1 |
ENST00000356450.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00149985.hg.1 |
ENST00000376006.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00149985.hg.1 |
NM_001304963.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00149985.hg.1 |
NM_153183.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00149985.hg.1 |
ENST00000356450.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00149985.hg.1 |
ENST00000376006.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00149985.hg.1 |
NM_001304963.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00149985.hg.1 |
NM_153183.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00149985.hg.1 |
TC0X00007261.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00009694.hg.1 |
ENST00000375992.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009694.hg.1 |
ENST00000375992.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009694.hg.1 |
ENST00000375992.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009694.hg.1 |
NM_018159.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009694.hg.1 |
NM_018159.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009694.hg.1 |
NM_018159.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007261.hg.1 |
ENST00000356450.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007261.hg.1 |
ENST00000356450.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007261.hg.1 |
ENST00000356450.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007261.hg.1 |
ENST00000356450.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007261.hg.1 |
ENST00000376006.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007261.hg.1 |
ENST00000376006.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007261.hg.1 |
ENST00000376006.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007261.hg.1 |
ENST00000376006.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007261.hg.1 |
NM_001304963.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007261.hg.1 |
NM_001304963.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007261.hg.1 |
NM_001304963.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007261.hg.1 |
NM_001304963.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007261.hg.1 |
NM_153183.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007261.hg.1 |
NM_153183.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007261.hg.1 |
NM_153183.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007261.hg.1 |
NM_153183.3 |
Refseq |