Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MDN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_014611.2 | NP_055426.1 | 99 - 100 | 16575 | 80 | |
XM_005248700.3 | XP_005248757.1 | 60 - 61 | 10507 | 80 | ||
XM_006715405.3 | XP_006715468.1 | 63 - 64 | 12053 | 80 | ||
XM_011535635.2 | XP_011533937.1 | 99 - 100 | 16380 | 80 | ||
GenBank mRNA | AB002299.2 | - | 53 - 54 | 9231 | 80 | |
AF503925.1 | - | 99 - 100 | 16504 | 80 | ||
AK027048.1 | - | 3 - 4 | 315 | 80 | ||
BC020432.1 | - | 5 - 6 | 558 | 80 | ||
BC036888.1 | - | 4 - 5 | 415 | 80 | ||
CR627399.1 | - | 45 - 46 | 8030 | 80 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |