Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012169.hg.1 |
ENST00000465434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012169.hg.1 |
ENST00000465434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010418.hg.1 |
ENST00000587474.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015173.hg.1 |
ENST00000448028.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011151.hg.1 |
ENST00000294189.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011151.hg.1 |
ENST00000294189.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011151.hg.1 |
ENST00000294189.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011151.hg.1 |
ENST00000294189.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011151.hg.1 |
ENST00000294189.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011151.hg.1 |
ENST00000294189.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011151.hg.1 |
ENST00000466397.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011151.hg.1 |
ENST00000466397.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011151.hg.1 |
ENST00000466397.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011151.hg.1 |
ENST00000466397.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011151.hg.1 |
ENST00000466397.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011151.hg.1 |
ENST00000466397.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011151.hg.1 |
ENST00000466397.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011151.hg.1 |
ENST00000475248.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011151.hg.1 |
ENST00000475248.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011151.hg.1 |
ENST00000475248.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011151.hg.1 |
ENST00000475248.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011151.hg.1 |
ENST00000475248.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011151.hg.1 |
ENST00000479017.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011151.hg.1 |
ENST00000479017.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011151.hg.1 |
ENST00000479017.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011151.hg.1 |
ENST00000479017.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011151.hg.1 |
ENST00000479017.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011151.hg.1 |
ENST00000479017.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011151.hg.1 |
ENST00000480306.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011151.hg.1 |
ENST00000480306.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011151.hg.1 |
ENST00000480306.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011151.hg.1 |
ENST00000480306.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011151.hg.1 |
ENST00000480306.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011151.hg.1 |
ENST00000480306.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011151.hg.1 |
ENST00000481629.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011151.hg.1 |
ENST00000481629.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011151.hg.1 |
ENST00000481629.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011151.hg.1 |
ENST00000481629.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011151.hg.1 |
ENST00000481629.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011151.hg.1 |
ENST00000481629.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011151.hg.1 |
ENST00000481629.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011151.hg.1 |
ENST00000486565.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011151.hg.1 |
ENST00000486565.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011151.hg.1 |
ENST00000486565.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011151.hg.1 |
ENST00000486565.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011151.hg.1 |
ENST00000486565.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011151.hg.1 |
ENST00000492277.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011151.hg.1 |
ENST00000492277.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011151.hg.1 |
ENST00000492277.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011151.hg.1 |
ENST00000492277.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011151.hg.1 |
ENST00000495383.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011151.hg.1 |
ENST00000495383.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011151.hg.1 |
ENST00000495383.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011151.hg.1 |
ENST00000495383.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011151.hg.1 |
ENST00000495383.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011151.hg.1 |
ENST00000495383.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011151.hg.1 |
ENST00000495383.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011151.hg.1 |
NM_000992.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011151.hg.1 |
NM_000992.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011151.hg.1 |
NM_000992.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011151.hg.1 |
NM_000992.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011151.hg.1 |
NM_000992.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011151.hg.1 |
NM_000992.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006988.hg.1 |
ENST00000426134.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006988.hg.1 |
ENST00000426134.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010989.hg.1 |
ENST00000446953.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010989.hg.1 |
ENST00000446953.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010418.hg.1 |
ENST00000587474.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010989.hg.1 |
ENST00000446953.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010989.hg.1 |
ENST00000446953.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015173.hg.1 |
ENST00000448028.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006988.hg.1 |
ENST00000426134.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006988.hg.1 |
ENST00000426134.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012169.hg.1 |
ENST00000465434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012169.hg.1 |
ENST00000465434.1 |
Ensembl |