Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04004615.hg.1 |
TC04000327.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04004615.hg.1 |
ENST00000257287.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04004615.hg.1 |
NM_025009.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
ENST00000257287.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
ENST00000257287.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
ENST00000257287.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
ENST00000257287.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
ENST00000257287.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
ENST00000257287.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
ENST00000257287.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
ENST00000257287.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
ENST00000257287.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
ENST00000257287.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
ENST00000257287.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
ENST00000257287.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
ENST00000257287.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
ENST00000257287.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
ENST00000257287.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
ENST00000257287.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
ENST00000257287.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
ENST00000257287.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
ENST00000257287.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
ENST00000257287.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
ENST00000257287.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
ENST00000257287.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
ENST00000257287.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
ENST00000257287.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
ENST00000257287.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
ENST00000257287.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
ENST00000257287.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
ENST00000257287.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
ENST00000257287.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
ENST00000257287.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
ENST00000422247.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
ENST00000422247.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
ENST00000422247.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
ENST00000422247.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
ENST00000422247.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
ENST00000422247.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
ENST00000422247.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
ENST00000422247.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
ENST00000422247.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
ENST00000422247.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
ENST00000422247.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
ENST00000506202.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
ENST00000506202.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
ENST00000506202.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
ENST00000506202.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
ENST00000506202.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
ENST00000506202.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
ENST00000506202.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
ENST00000506202.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
ENST00000506202.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
ENST00000506202.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
ENST00000506202.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
ENST00000506202.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
ENST00000506202.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
ENST00000506202.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
ENST00000506202.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
ENST00000506202.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
ENST00000506202.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
ENST00000506202.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
ENST00000506202.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
ENST00000506202.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
ENST00000506202.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
ENST00000506809.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
ENST00000506809.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
ENST00000506809.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
ENST00000506809.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
ENST00000506809.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
ENST00000515081.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
ENST00000515081.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
ENST00000515081.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
ENST00000515081.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
ENST00000515081.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
ENST00000515081.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
NM_025009.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
NM_025009.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
NM_025009.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
NM_025009.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
NM_025009.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
NM_025009.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
NM_025009.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
NM_025009.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
NM_025009.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
NM_025009.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
NM_025009.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
NM_025009.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
NM_025009.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
NM_025009.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
NM_025009.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
NM_025009.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
NM_025009.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
NM_025009.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
NM_025009.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
NM_025009.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
NM_025009.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
NM_025009.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
NM_025009.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
NM_025009.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
NM_025009.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
NM_025009.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
NM_025009.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
NM_025009.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
NM_025009.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
NM_025009.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
NM_025009.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
NM_025009.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007580.hg.1 |
NM_025009.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
ENST00000257287.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
ENST00000257287.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
ENST00000257287.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
ENST00000257287.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
ENST00000257287.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
ENST00000257287.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
ENST00000257287.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
ENST00000257287.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
ENST00000257287.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
ENST00000257287.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
ENST00000257287.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
ENST00000257287.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
ENST00000257287.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
ENST00000257287.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
ENST00000257287.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
ENST00000257287.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
ENST00000257287.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
ENST00000257287.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
ENST00000257287.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
ENST00000257287.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
ENST00000257287.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
ENST00000257287.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
ENST00000257287.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
ENST00000257287.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
ENST00000257287.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
ENST00000257287.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
ENST00000257287.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
ENST00000257287.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
ENST00000257287.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
ENST00000257287.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
ENST00000422247.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
ENST00000422247.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
ENST00000422247.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
ENST00000422247.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
ENST00000422247.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
ENST00000422247.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
ENST00000422247.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
ENST00000422247.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
ENST00000422247.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
ENST00000422247.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
ENST00000422247.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
ENST00000506202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
ENST00000506202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
ENST00000506202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
ENST00000506202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
ENST00000506202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
ENST00000506202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
ENST00000506202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
ENST00000506202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
ENST00000506202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
ENST00000506202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
ENST00000506202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
ENST00000506202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
ENST00000506202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
ENST00000506202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
ENST00000506202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
ENST00000506202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
ENST00000506202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
ENST00000506202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
ENST00000506202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
ENST00000506202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
ENST00000506202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
ENST00000506809.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
ENST00000506809.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
ENST00000506809.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
ENST00000506809.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
ENST00000506809.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
ENST00000515081.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
ENST00000515081.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
ENST00000515081.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
ENST00000515081.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
ENST00000515081.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
ENST00000515081.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
NM_025009.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
NM_025009.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
NM_025009.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
NM_025009.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
NM_025009.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
NM_025009.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
NM_025009.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
NM_025009.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
NM_025009.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
NM_025009.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
NM_025009.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
NM_025009.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
NM_025009.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
NM_025009.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
NM_025009.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
NM_025009.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
NM_025009.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
NM_025009.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
NM_025009.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
NM_025009.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
NM_025009.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
NM_025009.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
NM_025009.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
NM_025009.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
NM_025009.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
NM_025009.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
NM_025009.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
NM_025009.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
NM_025009.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
NM_025009.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
NM_025009.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
NM_025009.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007580.hg.1 |
NM_025009.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400151839.hg.1 |
ENST00000257287.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400151839.hg.1 |
ENST00000506202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400151839.hg.1 |
NM_025009.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400151839.hg.1 |
ENST00000257287.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400151839.hg.1 |
ENST00000506202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400151839.hg.1 |
NM_025009.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400151839.hg.1 |
TC0400007580.hg.1 |
Clariom D TC |