Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR15014977.hg.1 |
ENST00000352385.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15014977.hg.1 |
NM_020962.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15014977.hg.1 |
TC15001566.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009796.hg.1 |
ENST00000352385.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009796.hg.1 |
ENST00000352385.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009796.hg.1 |
ENST00000352385.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009796.hg.1 |
ENST00000352385.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009796.hg.1 |
ENST00000352385.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009796.hg.1 |
ENST00000352385.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009796.hg.1 |
ENST00000352385.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009796.hg.1 |
ENST00000352385.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009796.hg.1 |
ENST00000352385.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009796.hg.1 |
ENST00000352385.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009796.hg.1 |
ENST00000352385.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009796.hg.1 |
ENST00000352385.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009796.hg.1 |
ENST00000352385.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009796.hg.1 |
ENST00000352385.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009796.hg.1 |
ENST00000352385.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009796.hg.1 |
ENST00000352385.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009796.hg.1 |
ENST00000352385.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009796.hg.1 |
ENST00000352385.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009796.hg.1 |
ENST00000352385.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009796.hg.1 |
ENST00000352385.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009796.hg.1 |
ENST00000352385.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009796.hg.1 |
ENST00000352385.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009796.hg.1 |
ENST00000352385.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009796.hg.1 |
ENST00000352385.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009796.hg.1 |
ENST00000558048.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009796.hg.1 |
ENST00000558048.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009796.hg.1 |
ENST00000558048.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009796.hg.1 |
ENST00000558048.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009796.hg.1 |
ENST00000558048.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009796.hg.1 |
ENST00000558819.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009796.hg.1 |
ENST00000558819.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009796.hg.1 |
ENST00000559327.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009796.hg.1 |
ENST00000559327.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009796.hg.1 |
ENST00000559327.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009796.hg.1 |
ENST00000559327.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009796.hg.1 |
ENST00000559327.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009796.hg.1 |
ENST00000559327.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009796.hg.1 |
ENST00000559327.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009796.hg.1 |
ENST00000559327.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009796.hg.1 |
ENST00000559327.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009796.hg.1 |
ENST00000559327.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009796.hg.1 |
ENST00000559327.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009796.hg.1 |
ENST00000559327.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009796.hg.1 |
ENST00000559327.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009796.hg.1 |
ENST00000559327.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009796.hg.1 |
ENST00000559327.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009796.hg.1 |
ENST00000559327.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009796.hg.1 |
ENST00000559327.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009796.hg.1 |
ENST00000560319.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009796.hg.1 |
ENST00000560319.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009796.hg.1 |
ENST00000560319.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009796.hg.1 |
ENST00000560319.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009796.hg.1 |
ENST00000560319.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009796.hg.1 |
ENST00000561309.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009796.hg.1 |
ENST00000561309.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009796.hg.1 |
ENST00000561309.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009796.hg.1 |
NM_020962.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009796.hg.1 |
NM_020962.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009796.hg.1 |
NM_020962.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009796.hg.1 |
NM_020962.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009796.hg.1 |
NM_020962.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009796.hg.1 |
NM_020962.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009796.hg.1 |
NM_020962.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009796.hg.1 |
NM_020962.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009796.hg.1 |
NM_020962.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009796.hg.1 |
NM_020962.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009796.hg.1 |
NM_020962.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009796.hg.1 |
NM_020962.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009796.hg.1 |
NM_020962.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009796.hg.1 |
NM_020962.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009796.hg.1 |
NM_020962.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009796.hg.1 |
NM_020962.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009796.hg.1 |
NM_020962.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009796.hg.1 |
NM_020962.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009796.hg.1 |
NM_020962.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009796.hg.1 |
NM_020962.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009796.hg.1 |
NM_020962.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009796.hg.1 |
NM_020962.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009796.hg.1 |
NM_020962.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009796.hg.1 |
NM_020962.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500172428.hg.1 |
ENST00000352385.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500172428.hg.1 |
NM_020962.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500172428.hg.1 |
ENST00000352385.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500172428.hg.1 |
NM_020962.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500172428.hg.1 |
TC1500009796.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1500009796.hg.1 |
ENST00000352385.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009796.hg.1 |
ENST00000352385.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009796.hg.1 |
ENST00000352385.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009796.hg.1 |
ENST00000352385.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009796.hg.1 |
ENST00000352385.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009796.hg.1 |
ENST00000352385.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009796.hg.1 |
ENST00000352385.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009796.hg.1 |
ENST00000352385.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009796.hg.1 |
ENST00000352385.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009796.hg.1 |
ENST00000352385.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009796.hg.1 |
ENST00000352385.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009796.hg.1 |
ENST00000352385.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009796.hg.1 |
ENST00000352385.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009796.hg.1 |
ENST00000352385.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009796.hg.1 |
ENST00000352385.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009796.hg.1 |
ENST00000352385.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009796.hg.1 |
ENST00000352385.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009796.hg.1 |
ENST00000352385.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009796.hg.1 |
ENST00000352385.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009796.hg.1 |
ENST00000352385.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009796.hg.1 |
ENST00000352385.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009796.hg.1 |
ENST00000352385.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009796.hg.1 |
ENST00000352385.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009796.hg.1 |
ENST00000352385.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009796.hg.1 |
ENST00000558048.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009796.hg.1 |
ENST00000558048.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009796.hg.1 |
ENST00000558048.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009796.hg.1 |
ENST00000558048.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009796.hg.1 |
ENST00000558048.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009796.hg.1 |
ENST00000558819.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009796.hg.1 |
ENST00000558819.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009796.hg.1 |
ENST00000559327.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009796.hg.1 |
ENST00000559327.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009796.hg.1 |
ENST00000559327.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009796.hg.1 |
ENST00000559327.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009796.hg.1 |
ENST00000559327.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009796.hg.1 |
ENST00000559327.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009796.hg.1 |
ENST00000559327.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009796.hg.1 |
ENST00000559327.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009796.hg.1 |
ENST00000559327.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009796.hg.1 |
ENST00000559327.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009796.hg.1 |
ENST00000559327.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009796.hg.1 |
ENST00000559327.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009796.hg.1 |
ENST00000559327.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009796.hg.1 |
ENST00000559327.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009796.hg.1 |
ENST00000559327.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009796.hg.1 |
ENST00000559327.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009796.hg.1 |
ENST00000559327.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009796.hg.1 |
ENST00000560319.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009796.hg.1 |
ENST00000560319.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009796.hg.1 |
ENST00000560319.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009796.hg.1 |
ENST00000560319.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009796.hg.1 |
ENST00000560319.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009796.hg.1 |
ENST00000561309.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009796.hg.1 |
ENST00000561309.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009796.hg.1 |
ENST00000561309.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009796.hg.1 |
NM_020962.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009796.hg.1 |
NM_020962.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009796.hg.1 |
NM_020962.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009796.hg.1 |
NM_020962.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009796.hg.1 |
NM_020962.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009796.hg.1 |
NM_020962.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009796.hg.1 |
NM_020962.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009796.hg.1 |
NM_020962.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009796.hg.1 |
NM_020962.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009796.hg.1 |
NM_020962.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009796.hg.1 |
NM_020962.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009796.hg.1 |
NM_020962.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009796.hg.1 |
NM_020962.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009796.hg.1 |
NM_020962.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009796.hg.1 |
NM_020962.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009796.hg.1 |
NM_020962.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009796.hg.1 |
NM_020962.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009796.hg.1 |
NM_020962.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009796.hg.1 |
NM_020962.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009796.hg.1 |
NM_020962.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009796.hg.1 |
NM_020962.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009796.hg.1 |
NM_020962.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009796.hg.1 |
NM_020962.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009796.hg.1 |
NM_020962.2 |
Refseq |