Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01055501.hg.1 |
TC01003607.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01055501.hg.1 |
ENST00000367559.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01055501.hg.1 |
ENST00000444138.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01055501.hg.1 |
ENST00000539397.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01055501.hg.1 |
NM_021133.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016629.hg.1 |
ENST00000367559.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016629.hg.1 |
ENST00000367559.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016629.hg.1 |
ENST00000367559.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016629.hg.1 |
ENST00000367559.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016629.hg.1 |
ENST00000367559.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016629.hg.1 |
ENST00000367559.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016629.hg.1 |
ENST00000367559.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016629.hg.1 |
ENST00000367559.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016629.hg.1 |
ENST00000367559.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016629.hg.1 |
ENST00000367559.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016629.hg.1 |
ENST00000367559.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016629.hg.1 |
ENST00000539397.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016629.hg.1 |
ENST00000539397.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016629.hg.1 |
ENST00000539397.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016629.hg.1 |
ENST00000539397.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016629.hg.1 |
ENST00000539397.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016629.hg.1 |
ENST00000539397.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016629.hg.1 |
ENST00000539397.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016629.hg.1 |
ENST00000539397.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016629.hg.1 |
ENST00000539397.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016629.hg.1 |
NM_021133.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016629.hg.1 |
NM_021133.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016629.hg.1 |
NM_021133.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016629.hg.1 |
NM_021133.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016629.hg.1 |
NM_021133.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016629.hg.1 |
NM_021133.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016629.hg.1 |
NM_021133.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016629.hg.1 |
NM_021133.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016629.hg.1 |
NM_021133.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016629.hg.1 |
NM_021133.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016629.hg.1 |
NM_021133.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100226170.hg.1 |
ENST00000367559.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100226170.hg.1 |
ENST00000539397.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100226170.hg.1 |
NM_021133.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100226170.hg.1 |
ENST00000367559.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100226170.hg.1 |
ENST00000539397.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100226170.hg.1 |
NM_021133.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100226170.hg.1 |
TC0100016629.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100016629.hg.1 |
ENST00000367559.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016629.hg.1 |
ENST00000367559.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016629.hg.1 |
ENST00000367559.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016629.hg.1 |
ENST00000367559.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016629.hg.1 |
ENST00000367559.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016629.hg.1 |
ENST00000367559.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016629.hg.1 |
ENST00000367559.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016629.hg.1 |
ENST00000367559.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016629.hg.1 |
ENST00000367559.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016629.hg.1 |
ENST00000367559.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016629.hg.1 |
ENST00000367559.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016629.hg.1 |
ENST00000539397.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016629.hg.1 |
ENST00000539397.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016629.hg.1 |
ENST00000539397.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016629.hg.1 |
ENST00000539397.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016629.hg.1 |
ENST00000539397.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016629.hg.1 |
ENST00000539397.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016629.hg.1 |
ENST00000539397.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016629.hg.1 |
ENST00000539397.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016629.hg.1 |
ENST00000539397.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016629.hg.1 |
NM_021133.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016629.hg.1 |
NM_021133.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016629.hg.1 |
NM_021133.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016629.hg.1 |
NM_021133.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016629.hg.1 |
NM_021133.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016629.hg.1 |
NM_021133.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016629.hg.1 |
NM_021133.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016629.hg.1 |
NM_021133.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016629.hg.1 |
NM_021133.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016629.hg.1 |
NM_021133.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016629.hg.1 |
NM_021133.3 |
Refseq |