Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
ENST00000166345.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
ENST00000166345.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
ENST00000166345.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
ENST00000166345.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
ENST00000166345.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
ENST00000166345.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
ENST00000166345.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
ENST00000166345.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
ENST00000166345.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
ENST00000166345.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
ENST00000166345.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
ENST00000166345.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
ENST00000166345.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
ENST00000166345.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
ENST00000166345.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
ENST00000166345.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
ENST00000166345.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
ENST00000166345.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
ENST00000166345.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
ENST00000166345.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
ENST00000166345.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
ENST00000166345.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
ENST00000166345.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
ENST00000166345.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
ENST00000166345.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
ENST00000508430.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
ENST00000508430.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
ENST00000508430.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
ENST00000508456.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
ENST00000508456.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
ENST00000508456.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
ENST00000509210.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
ENST00000509210.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
ENST00000509210.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
ENST00000509210.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
ENST00000510412.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
ENST00000510412.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
ENST00000510412.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
ENST00000510412.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
ENST00000512024.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
ENST00000512024.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
ENST00000512024.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
ENST00000512024.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
ENST00000512024.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
ENST00000512024.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
ENST00000512024.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
ENST00000512024.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
ENST00000512024.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
ENST00000512024.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
ENST00000512024.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
ENST00000512024.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
ENST00000512024.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
ENST00000513435.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
ENST00000513435.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
ENST00000513435.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
ENST00000513435.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
ENST00000513435.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
ENST00000513435.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
ENST00000513435.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
ENST00000513435.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
ENST00000513435.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
NM_001166260.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
NM_001166260.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
NM_001166260.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
NM_001166260.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
NM_001166260.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
NM_001166260.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
NM_001166260.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
NM_001166260.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
NM_001166260.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
NM_001166260.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
NM_001166260.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
NM_001166260.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
NM_001166260.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
NM_001166260.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
NM_004237.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
NM_004237.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
NM_004237.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
NM_004237.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
NM_004237.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
NM_004237.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
NM_004237.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
NM_004237.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
NM_004237.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
NM_004237.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
NM_004237.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
NM_004237.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
NM_004237.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
NM_004237.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
NM_004237.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
NM_004237.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
NM_004237.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
NM_004237.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
NM_004237.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
NM_004237.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
NM_004237.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
NM_004237.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006472.hg.1 |
NM_004237.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05000257.hg.1 |
TC05000010.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05000257.hg.1 |
ENST00000166345.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05000257.hg.1 |
ENST00000354240.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05000257.hg.1 |
NM_001166260.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05000257.hg.1 |
NM_004237.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500144513.hg.1 |
ENST00000166345.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500144513.hg.1 |
ENST00000512024.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500144513.hg.1 |
ENST00000513435.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500144513.hg.1 |
NM_001166260.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500144513.hg.1 |
NM_004237.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500144513.hg.1 |
ENST00000166345.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500144513.hg.1 |
ENST00000512024.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500144513.hg.1 |
ENST00000513435.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500144513.hg.1 |
NM_001166260.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500144513.hg.1 |
NM_004237.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500144513.hg.1 |
TC0500006472.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
ENST00000166345.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
ENST00000166345.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
ENST00000166345.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
ENST00000166345.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
ENST00000166345.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
ENST00000166345.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
ENST00000166345.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
ENST00000166345.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
ENST00000166345.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
ENST00000166345.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
ENST00000166345.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
ENST00000166345.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
ENST00000166345.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
ENST00000166345.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
ENST00000166345.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
ENST00000166345.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
ENST00000166345.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
ENST00000166345.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
ENST00000166345.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
ENST00000166345.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
ENST00000166345.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
ENST00000166345.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
ENST00000166345.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
ENST00000166345.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
ENST00000166345.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
ENST00000508430.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
ENST00000508430.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
ENST00000508430.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
ENST00000508456.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
ENST00000508456.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
ENST00000508456.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
ENST00000509210.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
ENST00000509210.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
ENST00000509210.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
ENST00000509210.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
ENST00000510412.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
ENST00000510412.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
ENST00000510412.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
ENST00000510412.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
ENST00000512024.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
ENST00000512024.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
ENST00000512024.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
ENST00000512024.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
ENST00000512024.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
ENST00000512024.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
ENST00000512024.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
ENST00000512024.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
ENST00000512024.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
ENST00000512024.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
ENST00000512024.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
ENST00000512024.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
ENST00000512024.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
ENST00000513435.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
ENST00000513435.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
ENST00000513435.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
ENST00000513435.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
ENST00000513435.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
ENST00000513435.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
ENST00000513435.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
ENST00000513435.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
ENST00000513435.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
NM_001166260.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
NM_001166260.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
NM_001166260.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
NM_001166260.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
NM_001166260.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
NM_001166260.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
NM_001166260.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
NM_001166260.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
NM_001166260.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
NM_001166260.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
NM_001166260.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
NM_001166260.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
NM_001166260.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
NM_001166260.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
NM_004237.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
NM_004237.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
NM_004237.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
NM_004237.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
NM_004237.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
NM_004237.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
NM_004237.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
NM_004237.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
NM_004237.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
NM_004237.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
NM_004237.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
NM_004237.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
NM_004237.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
NM_004237.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
NM_004237.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
NM_004237.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
NM_004237.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
NM_004237.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
NM_004237.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
NM_004237.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
NM_004237.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
NM_004237.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006472.hg.1 |
NM_004237.3 |
Refseq |