Array Name |
Probe IDs |
Targets |
Target Type |
HumanHT-12 v4 Expression BeadChip |
ILMN_2102515 |
NM_001029891.2 |
human_transcriptome |
HumanHT-12 v4 Expression BeadChip |
ILMN_1691104 |
NM_001029891.2 |
human_transcriptome |
HumanHT-12 v4 Expression BeadChip |
ILMN_1682953 |
NM_001029891.2 |
human_transcriptome |
HumanHT-12 v4 Expression BeadChip |
ILMN_1682953 |
BG875904.1 |
human_transcriptome |
HumanHT-12 v4 Expression BeadChip |
ILMN_1706841 |
NM_001029891.2 |
human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip |
ILMN_1706841 |
NM_001029891.2 |
human_transcriptome |
HumanHT-12 V4.0 expression beadchip |
ILMN_1691104 |
NM_001029891.2 |
human_transcriptome |
HumanHT-12 V4.0 expression beadchip |
ILMN_1682953 |
NM_001029891.2 |
human_transcriptome |
HumanHT-12 V4.0 expression beadchip |
ILMN_1682953 |
BG875904.1 |
human_transcriptome |
HumanHT-12 V4.0 expression beadchip |
ILMN_1706841 |
NM_001029891.2 |
human_transcriptome |
HumanHT-12 V4.0 expression beadchip |
ILMN_2102515 |
NM_001029891.2 |
human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip |
ILMN_2102515 |
NM_001029891.2 |
human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip |
ILMN_2102515 |
NM_001029891.2 |
human_transcriptome |
Illumina HumanRef-8 v2 Expression BeadChip |
GI_139270-A |
- |
- |
Illumina Sentrix Human-6 v2 Expression BeadChip |
130092 |
BC042911.2 |
genbank_mrna_human |
Illumina Sentrix Human-6 v2 Expression BeadChip |
130092 |
BC061588.1 |
genbank_mrna_human |
Illumina Sentrix Human-6 v2 Expression BeadChip |
130092 |
BC073743.1 |
genbank_mrna_human |
Illumina Sentrix Human-6 v2 Expression BeadChip |
130092 |
CR456917.1 |
genbank_mrna_human |
Illumina Sentrix Human-6 v2 Expression BeadChip |
130092 |
L07769.1 |
genbank_mrna_human |
Illumina Sentrix Human-6 v2 Expression BeadChip |
130092 |
NM_001042507.2 |
refseq_mrna_human_NM |
Illumina Sentrix Human-6 v2 Expression BeadChip |
130092 |
NM_002307.2 |
refseq_mrna_human_NM |
Illumina Sentrix Human-6 v2 Expression BeadChip |
130092 |
U06643.1 |
genbank_mrna_human |
Illumina Sentrix Human-6 v2 Expression BeadChip |
130092 |
XM_001128786.1 |
refseq_mrna_human_XM |
Illumina Sentrix Human-6 v2 Expression BeadChip |
130092 |
XM_001133982.1 |
refseq_mrna_human_XM |
Illumina HumanRef-6 v2.0 expression beadchip |
ILMN_139270 |
- |
- |
Illumina HumanRef-8 v3.0 expression beadchip |
ILMN_2102515 |
NM_001029891.2 |
human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip |
ILMN_2102515 |
NM_001029891.2 |
human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip |
ILMN_1706841 |
NM_001029891.2 |
human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip |
ILMN_1682953 |
NM_001029891.2 |
human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip |
ILMN_1682953 |
BG875904.1 |
human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip |
ILMN_1706841 |
NM_001029891.2 |
human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip |
ILMN_2102515 |
NM_001029891.2 |
human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip |
ILMN_1691104 |
NM_001029891.2 |
human_transcriptome |
Illumina HumanWG-6_V2_0_R2 |
ILMN_1706841 |
NM_001029891.2 |
human_transcriptome |
Illumina HumanWG-6_V2_0_R2 |
ILMN_1691104 |
NM_001029891.2 |
human_transcriptome |
Illumina HumanWG-6_V2_0_R2 |
ILMN_1682953 |
NM_001029891.2 |
human_transcriptome |
Illumina HumanWG-6_V2_0_R2 |
ILMN_1682953 |
BG875904.1 |
human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 |
ILMN_2102515 |
NM_001029891.2 |
human_transcriptome |