Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: F8A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012151.3 | NP_036283.2 | 1 | 1659 | 88 | |
GenBank mRNA | BC039693.1 | - | 1 | 1645 | 5214628 | 88 |
BC071963.1 | - | 1 | 1649 | 5477018 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens 1700 Inventoried Both primers and probe map within a single exon |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P11262 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P11262 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 38777 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 36691 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 49547 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 36658 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 18063 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 52638 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 18236 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 37481 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 22709 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 60751 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 58837 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3242234 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P11262 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_19_P00323232 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_23_P11262 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_23_P11262 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P12038581 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P12038573 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P12038579 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P12038578 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P12038580 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P12038582 | - | - |
Agilent Human ENCODE ChIP-onChip A) | A_17_P12038579 | - | - |
Agilent Human ENCODE ChIP-onChip A) | A_17_P12038595 | - | - |
Agilent Human ENCODE ChIP-onChip A) | A_17_P12038582 | - | - |
Agilent Human ENCODE ChIP-onChip A) | A_17_P12038588 | - | - |
Agilent Human ENCODE ChIP-onChip A) | A_17_P12038589 | - | - |
Agilent Human ENCODE ChIP-onChip A) | A_17_P12038593 | - | - |
Agilent Human ENCODE ChIP-onChip A) | A_17_P12038584 | - | - |
Agilent Human ENCODE ChIP-onChip A) | A_17_P12038574 | - | - |
Agilent Human ENCODE ChIP-onChip A) | A_17_P12038590 | - | - |
Agilent Human ENCODE ChIP-onChip A) | A_17_P12038580 | - | - |
Agilent Human ENCODE ChIP-onChip A) | A_17_P12038581 | - | - |
Agilent Human ENCODE ChIP-onChip A) | A_17_P12038578 | - | - |
Agilent Human ENCODE ChIP-onChip A) | A_17_P12038583 | - | - |
Agilent Human ENCODE ChIP-onChip A) | A_17_P12038586 | - | - |
Agilent Human ENCODE ChIP-onChip A) | A_17_P12038592 | - | - |
Agilent Human ENCODE ChIP-onChip A) | A_17_P12038591 | - | - |
Agilent Human ENCODE ChIP-onChip A) | A_17_P12038573 | - | - |
Agilent Human ENCODE ChIP-onChip A) | A_17_P12038594 | - | - |
Agilent Human ENCODE ChIP-on-Chip Microarray 244K (G4495A) | A_17_P12038583 | - | - |
Agilent Human ENCODE ChIP-on-Chip Microarray 244K (G4495A) | A_17_P12038582 | - | - |
Agilent Human ENCODE ChIP-on-Chip Microarray 244K (G4495A) | A_17_P12038574 | - | - |
Agilent Human ENCODE ChIP-on-Chip Microarray 244K (G4495A) | A_17_P12038589 | - | - |
Agilent Human ENCODE ChIP-on-Chip Microarray 244K (G4495A) | A_17_P12038580 | - | - |
Agilent Human ENCODE ChIP-on-Chip Microarray 244K (G4495A) | A_17_P17417357 | - | - |
Agilent Human ENCODE ChIP-on-Chip Microarray 244K (G4495A) | A_17_P12038592 | - | - |
Agilent Human ENCODE ChIP-on-Chip Microarray 244K (G4495A) | A_17_P12038576 | - | - |
Agilent Human ENCODE ChIP-on-Chip Microarray 244K (G4495A) | A_17_P12038579 | - | - |
Agilent Human ENCODE ChIP-on-Chip Microarray 244K (G4495A) | A_17_P12038587 | - | - |
Agilent Human ENCODE ChIP-on-Chip Microarray 244K (G4495A) | A_17_P12038581 | - | - |
Agilent Human ENCODE ChIP-on-Chip Microarray 244K (G4495A) | A_17_P12038578 | - | - |
Agilent Human ENCODE ChIP-on-Chip Microarray 244K (G4495A) | A_17_P12038577 | - | - |
Agilent Human ENCODE ChIP-on-Chip Microarray 244K (G4495A) | A_17_P12038573 | - | - |
Agilent Human ENCODE ChIP-on-Chip Microarray 244K (G4495A) | A_17_P12038588 | - | - |
Agilent Human ENCODE ChIP-on-Chip Microarray 244K (G4495A) | A_17_P12038584 | - | - |
Agilent Human ENCODE ChIP-on-Chip Microarray 244K (G4495A) | A_17_P12038591 | - | - |
Agilent Human ENCODE ChIP-on-Chip Microarray 244K (G4495A) | A_17_P12038575 | - | - |
Agilent Human ENCODE ChIP-on-Chip Microarray 244K (G4495A) | A_17_P12038586 | - | - |
Agilent Human ENCODE ChIP-on-Chip Microarray 244K (G4495A) | A_17_P12038590 | - | - |
Agilent Human ENCODE ChIP-on-Chip Microarray 244K (G4495A) | A_17_P12038585 | - | - |
Agilent Human ENCODE ChIP-on-Chip Microarray 244K (G4495A) | A_17_P12038595 | - | - |
Agilent Human ENCODE ChIP-on-Chip Microarray 244K (G4495A) | A_17_P12038594 | - | - |
Agilent Human ENCODE ChIP-on-Chip Microarray 244K (G4495A) | A_17_P17417356 | - | - |
Agilent Human ENCODE ChIP-on-Chip Microarray 244K (G4495A) | A_17_P12038593 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P11262 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P12039902 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | 8171030 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8171031 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8171000 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8170999 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8176246 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8176247 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8176245 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8171029 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8170998 | - | - |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011242.hg.1 | ENST00000408263.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008860.hg.1 | ENST00000610495.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008860.hg.1 | ENST00000610495.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008860.hg.1 | NM_001007523.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008860.hg.1 | NM_001007524.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008860.hg.1 | NM_012151.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008860.hg.1 | NM_012151.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011227.hg.1 | ENST00000408606.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011227.hg.1 | NR_036049.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011227.hg.1 | NR_036259.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011227.hg.1 | NR_036260.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011244.hg.1 | ENST00000622749.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011244.hg.1 | ENST00000622749.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00011244.hg.1 | ENST00000622749.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008877.hg.1 | ENST00000408676.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X010610.hg.1 | TC0X000781.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR0X010610.hg.1 | ENST00000369446.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X010610.hg.1 | NM_012151.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00011227.hg.1 | ENST00000408606.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011227.hg.1 | NR_036049.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00011227.hg.1 | NR_036259.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00011227.hg.1 | NR_036260.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00011244.hg.1 | ENST00000622749.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011244.hg.1 | ENST00000622749.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011244.hg.1 | ENST00000622749.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00160780.hg.1 | ENST00000610495.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00160780.hg.1 | NM_012151.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00160780.hg.1 | ENST00000610495.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00160780.hg.1 | NM_012151.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00160780.hg.1 | TC0X00008860.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0X00008860.hg.1 | ENST00000610495.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008860.hg.1 | ENST00000610495.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008860.hg.1 | NM_001007523.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008860.hg.1 | NM_001007524.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008860.hg.1 | NM_012151.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008860.hg.1 | NM_012151.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008877.hg.1 | ENST00000408676.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00011242.hg.1 | ENST00000408263.1 | Ensembl |
Human Genome U133 Plus 2.0 Array | 203274_at | - | - |
Human Genome U133 Plus 2.0 Array | 203274_PM_at | - | - |
Human Genome U133A 2.0 Array | 203274_PM_at | - | - |
Human Genome U133A 2.0 Array | 203274_at | - | - |
AffymetrixPathwork Diagnostics Pathchip | pw565111 | - | - |
Human Genome U95 Set | 37907_at | - | - |
Human Genome U95 Set | 37907_PM_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 203274_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 203274_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 203274_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 203274_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 203274_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 203274_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 203274_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 203274_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 203274_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 203274_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 203274_at4 | - | - |
Human Genome U133 Set | 203274_at | - | - |
Human Genome U133 Set | 203274_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A041789 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A041788 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A041789 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A041788 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 194782 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_1790249 | BC039693.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1790249 | BC071963.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1790249 | NM_012151.3 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1790249 | BC039693.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1790249 | BC071963.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1790249 | NM_012151.3 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1790249 | BC039693.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1790249 | BC071963.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1790249 | NM_012151.3 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1790249 | BC039693.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1790249 | BC071963.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1790249 | NM_012151.3 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1790249 | BC039693.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1790249 | BC071963.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1790249 | NM_012151.3 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_12056462-S | CR623398.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_12056462-S | CR624283.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_12056462-S | CR621827.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_12056462-S | CR603094.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_12056462-S | CR615103.1 | human_transcriptome |
Illumina HumanRef-8 v2 Expression BeadChip | GI_23293-S | - | - |
Illumina human-6 v1.0 expression beadchip | 0004480519 | - | - |
Illumina humanRef-8 v1.0 expression beadchip | 0004480519 | - | - |
Illumina human-6 v2.0 expression beadchip | ILMN_1727984 | NM_001007523.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1727984 | NM_001007524.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1727984 | CR623398.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1727984 | CR624283.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1727984 | CR621827.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1790249 | BC039693.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1790249 | BC071963.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1790249 | NM_012151.3 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1727984 | NM_001007523.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1727984 | NM_001007524.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1727984 | CR623398.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1727984 | CR624283.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1727984 | CR621827.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1790249 | BC039693.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1790249 | BC071963.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1790249 | NM_012151.3 | human_transcriptome |
Illumina Sentrix Human-6 v2 Expression BeadChip | ILMN_23293 | - | - |
Illumina HumanRef-6 v2.0 expression beadchip | ILMN_23293 | - | - |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1790249 | BC039693.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1790249 | BC071963.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1790249 | NM_012151.3 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1790249 | BC039693.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1790249 | BC071963.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1790249 | NM_012151.3 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1790249 | BC039693.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1790249 | BC071963.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1790249 | NM_012151.3 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1790249 | BC039693.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1790249 | BC071963.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1790249 | NM_012151.3 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1790249 | BC039693.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1790249 | BC071963.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1790249 | NM_012151.3 | human_transcriptome |