Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19023746.hg.1 |
ENST00000376959.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19023746.hg.1 |
ENST00000391818.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19023746.hg.1 |
ENST00000443843.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19023746.hg.1 |
ENST00000474951.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19023746.hg.1 |
ENST00000477616.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19023746.hg.1 |
NM_004977.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19023746.hg.1 |
TC19001744.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19023748.hg.1 |
ENST00000376959.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19023748.hg.1 |
ENST00000443843.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19023748.hg.1 |
ENST00000477616.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19023748.hg.1 |
NM_004977.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19023748.hg.1 |
TC19001744.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011194.hg.1 |
ENST00000376959.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011194.hg.1 |
ENST00000376959.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011194.hg.1 |
ENST00000376959.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011194.hg.1 |
ENST00000376959.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011194.hg.1 |
ENST00000376959.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011194.hg.1 |
ENST00000376959.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011194.hg.1 |
ENST00000474951.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011194.hg.1 |
ENST00000474951.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011194.hg.1 |
ENST00000474951.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011194.hg.1 |
ENST00000474951.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011194.hg.1 |
ENST00000477616.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011194.hg.1 |
ENST00000477616.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011194.hg.1 |
ENST00000477616.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011194.hg.1 |
ENST00000477616.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011194.hg.1 |
ENST00000477616.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011194.hg.1 |
ENST00000477616.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011194.hg.1 |
ENST00000477616.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011194.hg.1 |
ENST00000477616.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011194.hg.1 |
NM_004977.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011194.hg.1 |
NM_004977.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011194.hg.1 |
NM_004977.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011194.hg.1 |
NM_004977.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011194.hg.1 |
NM_004977.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011194.hg.1 |
NM_004977.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011194.hg.1 |
NM_004977.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011194.hg.1 |
NM_004977.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011194.hg.1 |
NR_110912.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011194.hg.1 |
NR_110912.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011194.hg.1 |
NR_110912.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011194.hg.1 |
NR_110912.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19023747.hg.1 |
ENST00000376959.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19023747.hg.1 |
ENST00000443843.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19023747.hg.1 |
ENST00000477616.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19023747.hg.1 |
NM_004977.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19023747.hg.1 |
TC19001744.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19023749.hg.1 |
ENST00000443843.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19023749.hg.1 |
ENST00000477616.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19023749.hg.1 |
NM_004977.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19023749.hg.1 |
TC19001744.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1900193991.hg.1 |
ENST00000376959.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900193991.hg.1 |
ENST00000474951.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900193991.hg.1 |
ENST00000477616.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900193991.hg.1 |
NM_004977.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900193991.hg.1 |
NR_110912.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900193991.hg.1 |
TC1900011194.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900193991.hg.1 |
ENST00000376959.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900193991.hg.1 |
ENST00000474951.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900193991.hg.1 |
ENST00000477616.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900193991.hg.1 |
NM_004977.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900193991.hg.1 |
NR_110912.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900193995.hg.1 |
ENST00000477616.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900193995.hg.1 |
NM_004977.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900193995.hg.1 |
TC1900011194.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900193995.hg.1 |
ENST00000477616.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900193995.hg.1 |
NM_004977.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900193992.hg.1 |
ENST00000376959.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900193992.hg.1 |
ENST00000477616.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900193992.hg.1 |
NM_004977.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900193992.hg.1 |
TC1900011194.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900193992.hg.1 |
ENST00000376959.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900193992.hg.1 |
ENST00000477616.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900193992.hg.1 |
NM_004977.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900193993.hg.1 |
ENST00000376959.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900193993.hg.1 |
ENST00000477616.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900193993.hg.1 |
NM_004977.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900193993.hg.1 |
TC1900011194.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900193993.hg.1 |
ENST00000376959.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900193993.hg.1 |
ENST00000477616.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900193993.hg.1 |
NM_004977.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900193994.hg.1 |
ENST00000376959.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900193994.hg.1 |
ENST00000477616.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900193994.hg.1 |
NM_004977.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900193994.hg.1 |
TC1900011194.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900193994.hg.1 |
ENST00000376959.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900193994.hg.1 |
ENST00000477616.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900193994.hg.1 |
NM_004977.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011194.hg.1 |
ENST00000376959.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011194.hg.1 |
ENST00000376959.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011194.hg.1 |
ENST00000376959.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011194.hg.1 |
ENST00000376959.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011194.hg.1 |
ENST00000376959.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011194.hg.1 |
ENST00000376959.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011194.hg.1 |
ENST00000474951.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011194.hg.1 |
ENST00000474951.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011194.hg.1 |
ENST00000474951.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011194.hg.1 |
ENST00000474951.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011194.hg.1 |
ENST00000477616.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011194.hg.1 |
ENST00000477616.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011194.hg.1 |
ENST00000477616.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011194.hg.1 |
ENST00000477616.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011194.hg.1 |
ENST00000477616.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011194.hg.1 |
ENST00000477616.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011194.hg.1 |
ENST00000477616.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011194.hg.1 |
ENST00000477616.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011194.hg.1 |
NM_004977.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011194.hg.1 |
NM_004977.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011194.hg.1 |
NM_004977.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011194.hg.1 |
NM_004977.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011194.hg.1 |
NM_004977.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011194.hg.1 |
NM_004977.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011194.hg.1 |
NM_004977.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011194.hg.1 |
NM_004977.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011194.hg.1 |
NR_110912.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011194.hg.1 |
NR_110912.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011194.hg.1 |
NR_110912.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011194.hg.1 |
NR_110912.1 |
Refseq |