Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10001218.hg.1 |
ENST00000378372.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10001218.hg.1 |
NM_016299.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10001218.hg.1 |
TC10000112.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
ENST00000378372.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
ENST00000378372.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
ENST00000378372.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
ENST00000378372.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
ENST00000378372.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
ENST00000378372.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
ENST00000378372.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
ENST00000378372.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
ENST00000378372.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
ENST00000378372.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
ENST00000378372.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
ENST00000378372.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
ENST00000378372.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
ENST00000378372.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
ENST00000378372.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
ENST00000378372.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
ENST00000378372.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
ENST00000378372.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
ENST00000378372.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
ENST00000378372.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
ENST00000378372.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
ENST00000437161.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
ENST00000437161.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
ENST00000437161.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
ENST00000437161.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
ENST00000437161.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
ENST00000437161.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
ENST00000437161.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
ENST00000441647.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
ENST00000441647.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
ENST00000441647.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
ENST00000441647.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
ENST00000441647.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
ENST00000470430.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
ENST00000470430.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
ENST00000470430.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
ENST00000470430.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
ENST00000493178.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
ENST00000493178.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
ENST00000493178.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
ENST00000493178.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
ENST00000493178.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
ENST00000493178.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
ENST00000493863.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
ENST00000493863.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
ENST00000493863.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
ENST00000493863.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
ENST00000493863.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
ENST00000493863.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
ENST00000493863.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
ENST00000493863.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
NM_001278205.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
NM_001278205.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
NM_001278205.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
NM_001278205.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
NM_001278205.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
NM_001278205.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
NM_001278205.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
NM_001278205.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
NM_001278205.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
NM_001278205.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
NM_016299.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
NM_016299.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
NM_016299.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
NM_016299.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
NM_016299.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
NM_016299.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
NM_016299.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
NM_016299.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
NM_016299.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
NM_016299.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
NM_016299.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
NM_016299.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
NM_016299.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
NM_016299.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
NM_016299.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
NM_016299.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
NM_016299.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
NM_016299.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
NM_016299.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
NM_016299.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
NM_016299.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
NR_103464.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
NR_103464.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
NR_103464.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
NR_103464.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
NR_103464.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
NR_103464.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
NR_103464.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
NR_103464.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
NR_103464.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
NR_103464.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
NR_103464.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006858.hg.1 |
NR_103464.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10001219.hg.1 |
ENST00000378372.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10001219.hg.1 |
NM_016299.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10001219.hg.1 |
TC10000112.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1000146925.hg.1 |
ENST00000378372.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000146925.hg.1 |
ENST00000437161.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000146925.hg.1 |
ENST00000441647.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000146925.hg.1 |
ENST00000493178.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000146925.hg.1 |
ENST00000493863.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000146925.hg.1 |
NM_001278205.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000146925.hg.1 |
NM_016299.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000146925.hg.1 |
NR_103464.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000146925.hg.1 |
ENST00000378372.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000146925.hg.1 |
ENST00000437161.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000146925.hg.1 |
ENST00000441647.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000146925.hg.1 |
ENST00000493178.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000146925.hg.1 |
ENST00000493863.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000146925.hg.1 |
NM_001278205.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000146925.hg.1 |
NM_016299.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000146925.hg.1 |
NR_103464.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000146925.hg.1 |
TC1000006858.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
ENST00000378372.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
ENST00000378372.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
ENST00000378372.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
ENST00000378372.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
ENST00000378372.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
ENST00000378372.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
ENST00000378372.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
ENST00000378372.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
ENST00000378372.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
ENST00000378372.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
ENST00000378372.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
ENST00000378372.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
ENST00000378372.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
ENST00000378372.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
ENST00000378372.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
ENST00000378372.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
ENST00000378372.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
ENST00000378372.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
ENST00000378372.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
ENST00000378372.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
ENST00000378372.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
ENST00000437161.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
ENST00000437161.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
ENST00000437161.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
ENST00000437161.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
ENST00000437161.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
ENST00000437161.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
ENST00000437161.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
ENST00000441647.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
ENST00000441647.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
ENST00000441647.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
ENST00000441647.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
ENST00000441647.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
ENST00000470430.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
ENST00000470430.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
ENST00000470430.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
ENST00000470430.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
ENST00000493178.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
ENST00000493178.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
ENST00000493178.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
ENST00000493178.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
ENST00000493178.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
ENST00000493178.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
ENST00000493863.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
ENST00000493863.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
ENST00000493863.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
ENST00000493863.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
ENST00000493863.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
ENST00000493863.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
ENST00000493863.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
ENST00000493863.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
NM_001278205.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
NM_001278205.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
NM_001278205.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
NM_001278205.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
NM_001278205.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
NM_001278205.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
NM_001278205.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
NM_001278205.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
NM_001278205.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
NM_001278205.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
NM_016299.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
NM_016299.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
NM_016299.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
NM_016299.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
NM_016299.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
NM_016299.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
NM_016299.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
NM_016299.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
NM_016299.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
NM_016299.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
NM_016299.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
NM_016299.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
NM_016299.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
NM_016299.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
NM_016299.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
NM_016299.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
NM_016299.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
NM_016299.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
NM_016299.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
NM_016299.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
NM_016299.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
NR_103464.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
NR_103464.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
NR_103464.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
NR_103464.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
NR_103464.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
NR_103464.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
NR_103464.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
NR_103464.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
NR_103464.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
NR_103464.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
NR_103464.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006858.hg.1 |
NR_103464.1 |
Refseq |