Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01114010_g1
          See other CHRNB2 GE Assays ›
          Gene Symbol:
          CHRNB2
          Gene Name:
          cholinergic receptor nicotinic beta 2 subunit
          Gene Aliases:
          EFNL3, nAChRB2
          Chromosome Location:
          Chr.1: 154567781 - 154580026 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01114010_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CHRNB2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_000748.2 NP_000739.1 3 - 4 517 64
          XM_017000180.1 XP_016855669.1 3 - 4 499 64
          GenBank mRNA AK313470.1 - 3 - 4 286 64
          BC075040.2 - 3 - 4 282 30915357 64
          BC075041.2 - 3 - 4 282 30915573 64
          U62437.1 - 3 - 4 517 64
          X53179.1 - 3 - 4 419 64
          Y08415.1 - 3 - 4 253 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100010085.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01001279.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the XM_017000180.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CHRNB2

          Panther Classification:

          Molecular Function -

          ligand-gated ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          action potential
          conditioned taste aversion
          response to hypoxia
          ion transport
          calcium ion transport
          smooth muscle contraction
          signal transduction
          synaptic transmission, cholinergic
          neuromuscular synaptic transmission
          visual perception
          sensory perception of sound
          learning
          memory
          locomotory behavior
          associative learning
          visual learning
          regulation of dopamine secretion
          sensory perception of pain
          vestibulocochlear nerve development
          optic nerve morphogenesis
          lateral geniculate nucleus development
          central nervous system projection neuron axonogenesis
          positive regulation of B cell proliferation
          regulation of synaptic transmission, dopaminergic
          positive regulation of synaptic transmission, dopaminergic
          positive regulation of dopamine secretion
          response to nicotine
          behavioral response to nicotine
          social behavior
          regulation of dopamine metabolic process
          B cell activation
          response to cocaine
          regulation of circadian sleep/wake cycle, REM sleep
          regulation of circadian sleep/wake cycle, non-REM sleep
          response to ethanol
          negative regulation of action potential
          regulation of dendrite morphogenesis
          neurological system process
          cognition
          protein heterooligomerization
          membrane depolarization
          regulation of synapse assembly
          synaptic transmission involved in micturition
          cation transmembrane transport
          acetylcholine-activated cation-selective channel activity
          protein binding
          drug binding
          ligand-gated ion channel activity
          acetylcholine receptor activity
          acetylcholine binding
          protein heterodimerization activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0100010085.hg.1 ENST00000368476.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010085.hg.1 ENST00000368476.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010085.hg.1 ENST00000368476.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010085.hg.1 ENST00000368476.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010085.hg.1 ENST00000368476.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010085.hg.1 ENST00000368476.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010085.hg.1 ENST00000368476.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010085.hg.1 ENST00000368476.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010085.hg.1 ENST00000368476.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010085.hg.1 ENST00000368476.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010085.hg.1 ENST00000368476.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010085.hg.1 NM_000748.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010085.hg.1 NM_000748.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010085.hg.1 NM_000748.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010085.hg.1 NM_000748.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010085.hg.1 NM_000748.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010085.hg.1 NM_000748.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010085.hg.1 NM_000748.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010085.hg.1 NM_000748.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010085.hg.1 NM_000748.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010085.hg.1 NM_000748.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01019662.hg.1 TC01001279.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01019662.hg.1 ENST00000368476.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01019662.hg.1 NM_000748.2 Refseq
          Affymetrix Human Clariom D Assay TC0100010085.hg.1 ENST00000368476.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100010085.hg.1 ENST00000368476.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100010085.hg.1 ENST00000368476.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100010085.hg.1 ENST00000368476.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100010085.hg.1 ENST00000368476.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100010085.hg.1 ENST00000368476.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100010085.hg.1 ENST00000368476.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100010085.hg.1 ENST00000368476.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100010085.hg.1 ENST00000368476.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100010085.hg.1 ENST00000368476.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100010085.hg.1 ENST00000368476.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100010085.hg.1 NM_000748.2 Refseq
          Affymetrix Human Clariom D Assay TC0100010085.hg.1 NM_000748.2 Refseq
          Affymetrix Human Clariom D Assay TC0100010085.hg.1 NM_000748.2 Refseq
          Affymetrix Human Clariom D Assay TC0100010085.hg.1 NM_000748.2 Refseq
          Affymetrix Human Clariom D Assay TC0100010085.hg.1 NM_000748.2 Refseq
          Affymetrix Human Clariom D Assay TC0100010085.hg.1 NM_000748.2 Refseq
          Affymetrix Human Clariom D Assay TC0100010085.hg.1 NM_000748.2 Refseq
          Affymetrix Human Clariom D Assay TC0100010085.hg.1 NM_000748.2 Refseq
          Affymetrix Human Clariom D Assay TC0100010085.hg.1 NM_000748.2 Refseq
          Affymetrix Human Clariom D Assay TC0100010085.hg.1 NM_000748.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100173347.hg.1 ENST00000368476.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0100173347.hg.1 NM_000748.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100173347.hg.1 ENST00000368476.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0100173347.hg.1 NM_000748.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100173347.hg.1 TC0100010085.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline