Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNRPN | |||||||
---|---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF400500.1 | - | LIT2226 IC-SNURF-SNRPN 3 prime noncoding transcripts |
7 - 8 | 761 | 82 |
Set Membership: |
![]() ![]() ![]() ![]() |