Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR15018197.hg.1 |
ENST00000508732.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15018197.hg.1 |
ENST00000556899.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15018197.hg.1 |
NR_034095.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15018197.hg.1 |
TC15001917.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR15018198.hg.1 |
ENST00000508732.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15018198.hg.1 |
ENST00000556899.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15018198.hg.1 |
NR_034095.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15018198.hg.1 |
TC15001917.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010535.hg.1 |
ENST00000508732.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010535.hg.1 |
ENST00000508732.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010535.hg.1 |
ENST00000508732.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010535.hg.1 |
ENST00000508732.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010535.hg.1 |
ENST00000508732.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010535.hg.1 |
ENST00000508732.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010535.hg.1 |
ENST00000508732.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010535.hg.1 |
ENST00000508732.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010535.hg.1 |
ENST00000508732.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010535.hg.1 |
ENST00000508732.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010535.hg.1 |
ENST00000508732.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010535.hg.1 |
ENST00000508732.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010535.hg.1 |
ENST00000555332.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010535.hg.1 |
ENST00000555332.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010535.hg.1 |
ENST00000555332.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010535.hg.1 |
ENST00000555332.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010535.hg.1 |
ENST00000555332.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010535.hg.1 |
ENST00000555332.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010535.hg.1 |
ENST00000555332.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010535.hg.1 |
ENST00000555332.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010535.hg.1 |
ENST00000555332.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010535.hg.1 |
ENST00000555332.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010535.hg.1 |
ENST00000556899.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010535.hg.1 |
ENST00000556899.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010535.hg.1 |
ENST00000556899.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010535.hg.1 |
ENST00000556899.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010535.hg.1 |
ENST00000556899.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010535.hg.1 |
ENST00000611265.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010535.hg.1 |
ENST00000611265.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010535.hg.1 |
ENST00000611265.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010535.hg.1 |
ENST00000611265.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010535.hg.1 |
ENST00000611265.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010535.hg.1 |
ENST00000611265.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010535.hg.1 |
ENST00000611265.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010535.hg.1 |
ENST00000611265.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010535.hg.1 |
ENST00000611265.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010535.hg.1 |
ENST00000614853.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010535.hg.1 |
ENST00000614853.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010535.hg.1 |
ENST00000614853.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010535.hg.1 |
ENST00000614853.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010535.hg.1 |
ENST00000614853.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010535.hg.1 |
ENST00000616660.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010535.hg.1 |
ENST00000616660.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010535.hg.1 |
NR_034095.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010535.hg.1 |
NR_034095.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010535.hg.1 |
NR_034095.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010535.hg.1 |
NR_034095.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010535.hg.1 |
NR_034095.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010535.hg.1 |
NR_034095.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010535.hg.1 |
ENST00000508732.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010535.hg.1 |
ENST00000508732.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010535.hg.1 |
ENST00000508732.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010535.hg.1 |
ENST00000508732.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010535.hg.1 |
ENST00000508732.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010535.hg.1 |
ENST00000508732.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010535.hg.1 |
ENST00000508732.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010535.hg.1 |
ENST00000508732.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010535.hg.1 |
ENST00000508732.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010535.hg.1 |
ENST00000508732.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010535.hg.1 |
ENST00000508732.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010535.hg.1 |
ENST00000508732.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010535.hg.1 |
ENST00000555332.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010535.hg.1 |
ENST00000555332.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010535.hg.1 |
ENST00000555332.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010535.hg.1 |
ENST00000555332.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010535.hg.1 |
ENST00000555332.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010535.hg.1 |
ENST00000555332.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010535.hg.1 |
ENST00000555332.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010535.hg.1 |
ENST00000555332.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010535.hg.1 |
ENST00000555332.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010535.hg.1 |
ENST00000555332.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010535.hg.1 |
ENST00000556899.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010535.hg.1 |
ENST00000556899.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010535.hg.1 |
ENST00000556899.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010535.hg.1 |
ENST00000556899.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010535.hg.1 |
ENST00000556899.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010535.hg.1 |
ENST00000611265.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010535.hg.1 |
ENST00000611265.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010535.hg.1 |
ENST00000611265.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010535.hg.1 |
ENST00000611265.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010535.hg.1 |
ENST00000611265.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010535.hg.1 |
ENST00000611265.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010535.hg.1 |
ENST00000611265.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010535.hg.1 |
ENST00000611265.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010535.hg.1 |
ENST00000611265.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010535.hg.1 |
ENST00000614853.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010535.hg.1 |
ENST00000614853.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010535.hg.1 |
ENST00000614853.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010535.hg.1 |
ENST00000614853.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010535.hg.1 |
ENST00000614853.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010535.hg.1 |
ENST00000616660.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010535.hg.1 |
ENST00000616660.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010535.hg.1 |
NR_034095.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010535.hg.1 |
NR_034095.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010535.hg.1 |
NR_034095.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010535.hg.1 |
NR_034095.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010535.hg.1 |
NR_034095.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010535.hg.1 |
NR_034095.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500179252.hg.1 |
ENST00000508732.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500179252.hg.1 |
ENST00000508732.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500179252.hg.1 |
ENST00000556899.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500179252.hg.1 |
ENST00000611265.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500179252.hg.1 |
NR_034095.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500179252.hg.1 |
ENST00000508732.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500179252.hg.1 |
ENST00000508732.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500179252.hg.1 |
ENST00000556899.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500179252.hg.1 |
ENST00000611265.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500179252.hg.1 |
NR_034095.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500179252.hg.1 |
TC1500010535.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1500179253.hg.1 |
ENST00000508732.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500179253.hg.1 |
ENST00000508732.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500179253.hg.1 |
ENST00000556899.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500179253.hg.1 |
NR_034095.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500179253.hg.1 |
ENST00000508732.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500179253.hg.1 |
ENST00000508732.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500179253.hg.1 |
ENST00000556899.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500179253.hg.1 |
NR_034095.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500179253.hg.1 |
TC1500010535.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1500179250.hg.1 |
ENST00000508732.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500179250.hg.1 |
ENST00000508732.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500179250.hg.1 |
ENST00000556899.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500179250.hg.1 |
ENST00000611265.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500179250.hg.1 |
NR_034095.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500179250.hg.1 |
ENST00000508732.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500179250.hg.1 |
ENST00000508732.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500179250.hg.1 |
ENST00000556899.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500179250.hg.1 |
ENST00000611265.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500179250.hg.1 |
NR_034095.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500179250.hg.1 |
TC1500010535.hg.1 |
Clariom D TC |