Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000263646.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000263646.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000263646.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000263646.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000263646.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000263646.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000263646.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000263646.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000263646.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000263646.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000263646.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000317204.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000317204.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000317204.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000317204.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000317204.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000317204.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000317204.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000317204.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000317204.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000317204.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000317204.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000317204.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000317204.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000525159.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000525159.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000525159.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000525159.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000525159.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000525159.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000525159.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000525159.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000525159.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000525159.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000525159.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000525159.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000527085.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000527085.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000527638.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000527638.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000527638.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000527638.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000527746.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000527746.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000527746.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000527746.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000527746.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000527886.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000527886.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000527886.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000527886.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000527886.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000527886.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000527886.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000527886.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000527886.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000527886.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000527886.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000527886.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000527886.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000527938.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000527938.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000527938.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000527938.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000527938.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000527938.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000528719.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000528719.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000528719.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000530506.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000530506.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000530506.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000530506.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000530506.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000530506.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000530506.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000530506.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000530506.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000530541.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000530541.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000530541.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000530541.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000530541.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000530821.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000530821.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000532551.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000532551.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000532551.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
ENST00000532551.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
NM_019009.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
NM_019009.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
NM_019009.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
NM_019009.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
NM_019009.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
NM_019009.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
NM_019009.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
NM_019009.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
NM_019009.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
NM_019009.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
NM_019009.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
NM_019009.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009729.hg.1 |
NM_019009.3 |
Refseq |