Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR08022806.hg.1 |
TC08001729.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08022806.hg.1 |
ENST00000388913.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08022806.hg.1 |
NM_198488.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012160.hg.1 |
ENST00000388913.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012160.hg.1 |
ENST00000388913.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012160.hg.1 |
ENST00000388913.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012160.hg.1 |
ENST00000388913.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012160.hg.1 |
ENST00000388913.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012160.hg.1 |
ENST00000388913.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012160.hg.1 |
ENST00000388913.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012160.hg.1 |
ENST00000388913.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012160.hg.1 |
ENST00000388913.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012160.hg.1 |
ENST00000388913.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012160.hg.1 |
ENST00000388913.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012160.hg.1 |
ENST00000395103.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012160.hg.1 |
ENST00000395103.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012160.hg.1 |
ENST00000395103.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012160.hg.1 |
NM_198488.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012160.hg.1 |
NM_198488.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012160.hg.1 |
NM_198488.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012160.hg.1 |
NM_198488.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012160.hg.1 |
NM_198488.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012160.hg.1 |
NM_198488.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012160.hg.1 |
NM_198488.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012160.hg.1 |
NM_198488.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012160.hg.1 |
NM_198488.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012160.hg.1 |
NM_198488.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08022802.hg.1 |
TC08001729.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08022802.hg.1 |
ENST00000388913.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08022802.hg.1 |
NM_198488.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08022803.hg.1 |
TC08001729.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08022803.hg.1 |
ENST00000388913.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08022803.hg.1 |
NM_198488.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012161.hg.1 |
ENST00000583819.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012161.hg.1 |
NR_039810.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009213.hg.1 |
ENST00000435097.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009213.hg.1 |
ENST00000435097.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009213.hg.1 |
ENST00000435097.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009213.hg.1 |
ENST00000435097.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009213.hg.1 |
ENST00000435097.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009213.hg.1 |
ENST00000533004.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009213.hg.1 |
ENST00000533004.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009213.hg.1 |
ENST00000533004.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009213.hg.1 |
ENST00000533004.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009213.hg.1 |
ENST00000533004.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009213.hg.1 |
ENST00000533004.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009213.hg.1 |
ENST00000533004.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009213.hg.1 |
ENST00000533004.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009213.hg.1 |
ENST00000534398.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009213.hg.1 |
ENST00000534398.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009213.hg.1 |
ENST00000534398.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800182992.hg.1 |
ENST00000388913.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800182992.hg.1 |
NM_198488.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800182992.hg.1 |
ENST00000388913.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800182992.hg.1 |
NM_198488.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800182992.hg.1 |
TC0800012160.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0800012161.hg.1 |
ENST00000583819.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012161.hg.1 |
NR_039810.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012160.hg.1 |
ENST00000388913.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012160.hg.1 |
ENST00000388913.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012160.hg.1 |
ENST00000388913.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012160.hg.1 |
ENST00000388913.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012160.hg.1 |
ENST00000388913.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012160.hg.1 |
ENST00000388913.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012160.hg.1 |
ENST00000388913.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012160.hg.1 |
ENST00000388913.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012160.hg.1 |
ENST00000388913.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012160.hg.1 |
ENST00000388913.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012160.hg.1 |
ENST00000388913.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012160.hg.1 |
ENST00000395103.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012160.hg.1 |
ENST00000395103.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012160.hg.1 |
ENST00000395103.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012160.hg.1 |
NM_198488.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012160.hg.1 |
NM_198488.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012160.hg.1 |
NM_198488.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012160.hg.1 |
NM_198488.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012160.hg.1 |
NM_198488.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012160.hg.1 |
NM_198488.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012160.hg.1 |
NM_198488.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012160.hg.1 |
NM_198488.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012160.hg.1 |
NM_198488.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012160.hg.1 |
NM_198488.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800182991.hg.1 |
ENST00000388913.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800182991.hg.1 |
NM_198488.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800182991.hg.1 |
ENST00000388913.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800182991.hg.1 |
NM_198488.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800182991.hg.1 |
TC0800012160.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800182994.hg.1 |
ENST00000388913.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800182994.hg.1 |
NM_198488.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800182994.hg.1 |
ENST00000388913.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800182994.hg.1 |
NM_198488.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800182994.hg.1 |
TC0800012160.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0800009213.hg.1 |
ENST00000435097.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009213.hg.1 |
ENST00000435097.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009213.hg.1 |
ENST00000435097.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009213.hg.1 |
ENST00000435097.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009213.hg.1 |
ENST00000435097.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009213.hg.1 |
ENST00000533004.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009213.hg.1 |
ENST00000533004.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009213.hg.1 |
ENST00000533004.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009213.hg.1 |
ENST00000533004.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009213.hg.1 |
ENST00000533004.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009213.hg.1 |
ENST00000533004.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009213.hg.1 |
ENST00000533004.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009213.hg.1 |
ENST00000533004.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009213.hg.1 |
ENST00000534398.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009213.hg.1 |
ENST00000534398.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009213.hg.1 |
ENST00000534398.1 |
Ensembl |