Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | M13930.1 | - | 2 - 3 | 1963 | 105 |
Set Membership: |
![]() ![]() ![]() ![]() |