Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008259.hg.1 |
ENST00000391958.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008259.hg.1 |
ENST00000391958.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008259.hg.1 |
ENST00000391958.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008259.hg.1 |
ENST00000391958.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008259.hg.1 |
ENST00000391958.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008259.hg.1 |
ENST00000391958.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008259.hg.1 |
ENST00000391958.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008259.hg.1 |
ENST00000391958.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008259.hg.1 |
ENST00000391958.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008259.hg.1 |
ENST00000588489.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008259.hg.1 |
ENST00000588489.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008259.hg.1 |
ENST00000588489.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008259.hg.1 |
ENST00000588489.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008259.hg.1 |
ENST00000589522.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008259.hg.1 |
ENST00000589522.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008259.hg.1 |
ENST00000589522.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008259.hg.1 |
ENST00000589522.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008259.hg.1 |
ENST00000590668.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008259.hg.1 |
ENST00000590668.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008259.hg.1 |
ENST00000590668.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008259.hg.1 |
ENST00000590668.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008259.hg.1 |
ENST00000590668.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008259.hg.1 |
ENST00000592581.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008259.hg.1 |
ENST00000592581.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008259.hg.1 |
ENST00000592581.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008259.hg.1 |
ENST00000592581.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008259.hg.1 |
ENST00000592581.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008259.hg.1 |
ENST00000592581.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008259.hg.1 |
ENST00000592581.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008259.hg.1 |
ENST00000592581.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008259.hg.1 |
ENST00000592581.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008259.hg.1 |
NM_001207005.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008259.hg.1 |
NM_001207005.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008259.hg.1 |
NM_001207005.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008259.hg.1 |
NM_001207005.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008259.hg.1 |
NM_001207005.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008259.hg.1 |
NM_001207005.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008259.hg.1 |
NM_001207005.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008259.hg.1 |
NM_001207005.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008259.hg.1 |
NM_181756.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008259.hg.1 |
NM_181756.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008259.hg.1 |
NM_181756.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008259.hg.1 |
NM_181756.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008259.hg.1 |
NM_181756.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008259.hg.1 |
NM_181756.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008259.hg.1 |
NM_181756.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008259.hg.1 |
NM_181756.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008259.hg.1 |
NM_181756.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19008610.hg.1 |
ENST00000334152.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19008610.hg.1 |
NM_001207005.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19008610.hg.1 |
TC19000622.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19008617.hg.1 |
ENST00000334152.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19008617.hg.1 |
ENST00000391958.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19008617.hg.1 |
NM_001207005.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19008617.hg.1 |
NM_181756.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19008617.hg.1 |
TC19000622.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1900008259.hg.1 |
ENST00000391958.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008259.hg.1 |
ENST00000391958.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008259.hg.1 |
ENST00000391958.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008259.hg.1 |
ENST00000391958.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008259.hg.1 |
ENST00000391958.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008259.hg.1 |
ENST00000391958.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008259.hg.1 |
ENST00000391958.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008259.hg.1 |
ENST00000391958.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008259.hg.1 |
ENST00000391958.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008259.hg.1 |
ENST00000588489.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008259.hg.1 |
ENST00000588489.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008259.hg.1 |
ENST00000588489.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008259.hg.1 |
ENST00000588489.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008259.hg.1 |
ENST00000589522.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008259.hg.1 |
ENST00000589522.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008259.hg.1 |
ENST00000589522.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008259.hg.1 |
ENST00000589522.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008259.hg.1 |
ENST00000590668.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008259.hg.1 |
ENST00000590668.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008259.hg.1 |
ENST00000590668.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008259.hg.1 |
ENST00000590668.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008259.hg.1 |
ENST00000590668.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008259.hg.1 |
ENST00000592581.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008259.hg.1 |
ENST00000592581.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008259.hg.1 |
ENST00000592581.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008259.hg.1 |
ENST00000592581.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008259.hg.1 |
ENST00000592581.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008259.hg.1 |
ENST00000592581.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008259.hg.1 |
ENST00000592581.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008259.hg.1 |
ENST00000592581.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008259.hg.1 |
ENST00000592581.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008259.hg.1 |
NM_001207005.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008259.hg.1 |
NM_001207005.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008259.hg.1 |
NM_001207005.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008259.hg.1 |
NM_001207005.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008259.hg.1 |
NM_001207005.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008259.hg.1 |
NM_001207005.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008259.hg.1 |
NM_001207005.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008259.hg.1 |
NM_001207005.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008259.hg.1 |
NM_181756.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008259.hg.1 |
NM_181756.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008259.hg.1 |
NM_181756.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008259.hg.1 |
NM_181756.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008259.hg.1 |
NM_181756.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008259.hg.1 |
NM_181756.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008259.hg.1 |
NM_181756.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008259.hg.1 |
NM_181756.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008259.hg.1 |
NM_181756.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900163073.hg.1 |
ENST00000391958.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900163073.hg.1 |
ENST00000588489.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900163073.hg.1 |
ENST00000589522.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900163073.hg.1 |
ENST00000590668.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900163073.hg.1 |
ENST00000592581.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900163073.hg.1 |
NM_001207005.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900163073.hg.1 |
NM_181756.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900163073.hg.1 |
TC1900008259.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900163073.hg.1 |
ENST00000391958.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900163073.hg.1 |
ENST00000588489.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900163073.hg.1 |
ENST00000589522.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900163073.hg.1 |
ENST00000590668.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900163073.hg.1 |
ENST00000592581.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900163073.hg.1 |
NM_001207005.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900163073.hg.1 |
NM_181756.2 |
Refseq |