Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CT83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017978.3 | NP_001017978.1 | 1 - 2 | 249 | 80 | |
GenBank mRNA | AK000618.1 | - | 1 - 2 | 219 | 80 | |
AK026566.1 | - | 1 - 2 | 219 | 80 | ||
BC062223.1 | - | 1 - 2 | 196 | 6101827 | 80 | |
HY170569.1 | - | 1 - 2 | 250 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens Inventoried Probe spans exons |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11905000 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P17390005 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P17389998 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P17390003 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32635626 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P17390001 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32635632 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32635641 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32635633 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P17390007 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32635645 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P17389993 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32635642 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32635635 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11905010 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P17390000 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11905006 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P17390009 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32635625 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32635637 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P432352 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P432352 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 1740 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 8278 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_18_P17277799 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P432352 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_23_P432352 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32635642 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32635625 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17390001 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17390009 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17390003 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17390005 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32635639 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32635635 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32635633 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17390000 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11905006 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32635641 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32635645 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17389998 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17389993 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11905010 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32635626 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32635627 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17390007 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32635632 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11905000 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17390007 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32635627 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17389998 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32635625 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17390005 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17390003 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11905010 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11905006 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17390009 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32635633 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17390001 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17389993 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17390000 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32635639 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32635626 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32635641 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11905000 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32635635 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32635645 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32635632 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32635642 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_23_P432352 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P11904998 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P11905011 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P11905009 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P11905002 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P11905000 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P11905003 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P11905006 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P11905005 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P11905010 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P432352 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11905002 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11905000 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11905004 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11905010 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11905005 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11905009 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11904998 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | NM_001017978_at | - | - |
Affymetrix Human Gene 1.0 ST Array | 8174649 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8174651 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8174650 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8174648 | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 1559258_PM_a_at | - | - |
Affymetrix Human Genome U219 Array | 11732935_at | - | - |
Affymetrix Human Genome U219 Array | 11732935_PM_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | PSR0X017500.hg.1 | TC0X001299.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR0X017500.hg.1 | ENST00000371894.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X017500.hg.1 | NM_001017978.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010596.hg.1 | ENST00000371894.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010596.hg.1 | ENST00000371894.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010596.hg.1 | ENST00000371894.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010596.hg.1 | ENST00000371894.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010596.hg.1 | ENST00000371894.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010596.hg.1 | NM_001017978.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010596.hg.1 | NM_001017978.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010596.hg.1 | NM_001017978.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010596.hg.1 | NM_001017978.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010596.hg.1 | NM_001017978.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010596.hg.1 | NM_001017978.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR0X017501.hg.1 | TC0X001299.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR0X017501.hg.1 | ENST00000371894.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X017501.hg.1 | NM_001017978.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010599.hg.1 | ENST00000446495.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010599.hg.1 | ENST00000446495.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010596.hg.1 | ENST00000371894.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010596.hg.1 | ENST00000371894.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010596.hg.1 | ENST00000371894.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010596.hg.1 | ENST00000371894.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010596.hg.1 | ENST00000371894.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010596.hg.1 | NM_001017978.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010596.hg.1 | NM_001017978.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010596.hg.1 | NM_001017978.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010596.hg.1 | NM_001017978.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010596.hg.1 | NM_001017978.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010596.hg.1 | NM_001017978.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00172047.hg.1 | ENST00000371894.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00172047.hg.1 | NM_001017978.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00172047.hg.1 | ENST00000371894.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00172047.hg.1 | NM_001017978.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00172047.hg.1 | TC0X00010596.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0X00172046.hg.1 | ENST00000371894.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00172046.hg.1 | NM_001017978.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00172046.hg.1 | ENST00000371894.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00172046.hg.1 | NM_001017978.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00172046.hg.1 | TC0X00010596.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0X00172048.hg.1 | ENST00000371894.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00172048.hg.1 | NM_001017978.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00172048.hg.1 | ENST00000371894.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00172048.hg.1 | NM_001017978.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00172048.hg.1 | TC0X00010596.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0X00172050.hg.1 | ENST00000371894.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00172050.hg.1 | NM_001017978.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00172050.hg.1 | ENST00000371894.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00172050.hg.1 | NM_001017978.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00172050.hg.1 | TC0X00010596.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0X00172051.hg.1 | NM_001017978.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00172051.hg.1 | NM_001017978.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00172051.hg.1 | TC0X00010596.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0X00172049.hg.1 | ENST00000371894.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00172049.hg.1 | NM_001017978.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00172049.hg.1 | ENST00000371894.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00172049.hg.1 | NM_001017978.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00172049.hg.1 | TC0X00010596.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0X00010599.hg.1 | ENST00000446495.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010599.hg.1 | ENST00000446495.1 | Ensembl |
Human Genome U133 Plus 2.0 Array | 1559258_a_at | - | - |
Human Genome U133 Plus 2.0 Array | 1559258_PM_a_at | - | - |
Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) | NM_001017978_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1559258_a_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1559258_a_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1559258_a_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1559258_a_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1559258_a_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1559258_a_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1559258_a_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1559258_a_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1559258_a_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1559258_a_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1559258_a_at8 | - | - |
Human Genome U95 Set | 57298_PM_at | - | - |
Human Genome U95 Set | 57298_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A040305 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A040305 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_1668108 | AK000618.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1668108 | AK026566.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1668108 | BC062223.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1668108 | NM_001017978.2 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1668108 | AK000618.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1668108 | AK026566.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1668108 | BC062223.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1668108 | NM_001017978.2 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1668108 | AK000618.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1668108 | AK026566.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1668108 | BC062223.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1668108 | NM_001017978.2 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1668108 | AK000618.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1668108 | AK026566.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1668108 | BC062223.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1668108 | NM_001017978.2 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1668108 | AK000618.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1668108 | AK026566.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1668108 | BC062223.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1668108 | NM_001017978.2 | human_transcriptome |
Illumina HumanRef-8 v2 Expression BeadChip | GI_18870-S | - | - |
Illumina human-6 v2.0 expression beadchip | ILMN_1668108 | AK000618.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1668108 | AK026566.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1668108 | BC062223.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1668108 | NM_001017978.2 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1668108 | AK000618.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1668108 | AK026566.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1668108 | BC062223.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1668108 | NM_001017978.2 | human_transcriptome |
Illumina Sentrix Human-6 v2 Expression BeadChip | ILMN_18870 | - | - |
Illumina HumanRef-6 v2.0 expression beadchip | ILMN_18870 | - | - |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1668108 | AK000618.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1668108 | AK026566.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1668108 | BC062223.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1668108 | NM_001017978.2 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1668108 | AK000618.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1668108 | AK026566.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1668108 | BC062223.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1668108 | NM_001017978.2 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1668108 | AK000618.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1668108 | AK026566.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1668108 | BC062223.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1668108 | NM_001017978.2 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1668108 | AK000618.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1668108 | AK026566.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1668108 | BC062223.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1668108 | NM_001017978.2 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1668108 | AK000618.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1668108 | AK026566.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1668108 | BC062223.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1668108 | NM_001017978.2 | human_transcriptome |