Save up to 49% with protein expression workflow promo

Save now

Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Gene Expression
  • Search All
  • Aspire Free Products
  • Popular Product Areas
  • TaqMan Assays
  • Gene Expression
  • Microbe & ABR Detection
  • Copy Number Variation
  • Mutation Detection
  • miRNA
  • SNP Genotyping
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central
            This product has been added to your favorites list. Go to My Favorites

            System Message

            OKCancel
            LOADING ...
            • Home
            • › Search Tool
            • › Search Results
            • › Hs02513543_s1
            See other ALDH4A1 GE Assays ›
            Gene Symbol:
            ALDH4A1
            Gene Name:
            aldehyde dehydrogenase 4 family member A1
            Gene Aliases:
            ALDH4, P5CD, P5CDh
            Chromosome Location:
            Chr.1: 18871430 - 18902799 on Build GRCh38
            Species:
            Human
            Species Specific ID (Flybase ID):
            -
            Assay ID Hs02513543_s1
            Size
            Availability Made To Order
            Catalog # 4351372
            Price
            Your Price
            Online offer:
            Check your price ›
            • Genomic Map
            • Assay Details
            • More Information

            Genomic Map

            LOADING... Fetching data..
            ×
            Back To Top

            Assay Details

            Gene Symbol: ALDH4A1
            Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
            RefSeq NM_001161504.1 NP_001154976.1 15 2818 95
            NM_001319218.1 NP_001306147.1 14 2805 95
            NM_003748.3 NP_003739.2 15 2958 95
            GenBank mRNA BC007581.1 - 15 2688 3139944 95
            BC023600.2 - 15 2699 4548787 95
            BX649081.1 - 2 2061 95
            FJ462711.1 - 14 2543 95
            U24266.1 - 15 2710 95

            Back To Top

            More Information


            Additional Information:

            Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

            Set Membership:

            Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

            Matching Ambion siRNAs:

            Click here to find siRNAs for Gene ALDH4A1

            Panther Classification:

            Molecular Function -

            dehydrogenase oxidoreductase metabolite interconversion enzyme

            Gene Ontology Categories:

            Function(s) Process(es)

            glutamate biosynthetic process
            proline metabolic process
            proline biosynthetic process
            proline catabolic process
            proline catabolic process to glutamate
            4-hydroxyproline catabolic process
            oxidation-reduction process
            1-pyrroline-5-carboxylate dehydrogenase activity
            aldehyde dehydrogenase (NAD) activity
            electron carrier activity
            identical protein binding

            Back To Top

            Related Products

            • TaqMan® Fast Advanced Master Mix
            • SuperScript® VILO™ cDNA Synthesis Kit
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            • Price & Freight Policy
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • German Supply Chain Due Diligence Act
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Legal Notice
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            Germany flag icon
            Germany

            Your items have has been added!


            Host server : magellan-search-green-6ff95d844f-rlz7t:80/100.66.77.154:80.
            git-commit: f44aa9153aef922f8fad9a05e87b8aee811da128
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.28.0-Offline