Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09012654.hg.1 |
TC09000946.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09012654.hg.1 |
ENST00000380232.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09012654.hg.1 |
NM_002176.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009688.hg.1 |
ENST00000380232.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009688.hg.1 |
ENST00000380232.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009688.hg.1 |
ENST00000380232.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009688.hg.1 |
ENST00000380232.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009688.hg.1 |
NM_002176.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009688.hg.1 |
NM_002176.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009688.hg.1 |
NM_002176.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009688.hg.1 |
NM_002176.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900166290.hg.1 |
ENST00000380232.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900166290.hg.1 |
NM_002176.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900166290.hg.1 |
ENST00000380232.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900166290.hg.1 |
NM_002176.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900166290.hg.1 |
TC0900009688.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900009688.hg.1 |
ENST00000380232.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009688.hg.1 |
ENST00000380232.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009688.hg.1 |
ENST00000380232.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009688.hg.1 |
ENST00000380232.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009688.hg.1 |
NM_002176.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009688.hg.1 |
NM_002176.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009688.hg.1 |
NM_002176.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009688.hg.1 |
NM_002176.3 |
Refseq |