Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011212.hg.1 |
ENST00000306388.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011212.hg.1 |
ENST00000306388.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011212.hg.1 |
ENST00000306388.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011212.hg.1 |
ENST00000306388.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011212.hg.1 |
ENST00000306388.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011212.hg.1 |
ENST00000306388.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011212.hg.1 |
ENST00000306388.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011212.hg.1 |
ENST00000380377.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011212.hg.1 |
ENST00000380377.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011212.hg.1 |
ENST00000380377.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011212.hg.1 |
ENST00000380377.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011212.hg.1 |
ENST00000380377.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011212.hg.1 |
ENST00000380377.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011212.hg.1 |
ENST00000380377.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011212.hg.1 |
ENST00000517679.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011212.hg.1 |
ENST00000517679.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011212.hg.1 |
ENST00000517679.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011212.hg.1 |
ENST00000517679.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011212.hg.1 |
ENST00000517679.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011212.hg.1 |
ENST00000517881.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011212.hg.1 |
ENST00000517881.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011212.hg.1 |
ENST00000517881.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011212.hg.1 |
ENST00000517881.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011212.hg.1 |
ENST00000518218.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011212.hg.1 |
ENST00000518218.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011212.hg.1 |
ENST00000518218.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011212.hg.1 |
ENST00000518218.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011212.hg.1 |
ENST00000518218.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011212.hg.1 |
ENST00000518338.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011212.hg.1 |
ENST00000518338.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011212.hg.1 |
ENST00000518338.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011212.hg.1 |
ENST00000518338.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011212.hg.1 |
ENST00000518338.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011212.hg.1 |
ENST00000518338.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011212.hg.1 |
ENST00000520039.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011212.hg.1 |
ENST00000520039.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011212.hg.1 |
ENST00000520361.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011212.hg.1 |
ENST00000520361.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011212.hg.1 |
ENST00000520361.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011212.hg.1 |
ENST00000520361.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011212.hg.1 |
ENST00000522370.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011212.hg.1 |
ENST00000522370.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011212.hg.1 |
ENST00000522370.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011212.hg.1 |
ENST00000522370.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011212.hg.1 |
NM_001297738.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011212.hg.1 |
NM_001297738.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011212.hg.1 |
NM_001297738.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011212.hg.1 |
NM_001297738.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011212.hg.1 |
NM_001297738.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011212.hg.1 |
NM_001297738.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011212.hg.1 |
NM_001297738.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011212.hg.1 |
NM_001297738.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011212.hg.1 |
NM_001297738.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011212.hg.1 |
NM_001297740.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011212.hg.1 |
NM_001297740.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011212.hg.1 |
NM_001297740.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011212.hg.1 |
NM_001297740.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011212.hg.1 |
NM_001297740.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011212.hg.1 |
NM_001297740.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011212.hg.1 |
NM_004607.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011212.hg.1 |
NM_004607.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011212.hg.1 |
NM_004607.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011212.hg.1 |
NM_004607.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011212.hg.1 |
NM_004607.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011212.hg.1 |
NM_004607.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011212.hg.1 |
NM_004607.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05020698.hg.1 |
TC05001510.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05020698.hg.1 |
ENST00000306388.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05020698.hg.1 |
ENST00000380377.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05020698.hg.1 |
ENST00000520039.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05020698.hg.1 |
ENST00000520361.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05020698.hg.1 |
NM_004607.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05020703.hg.1 |
TC05001510.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05020703.hg.1 |
ENST00000380377.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05020703.hg.1 |
NM_004607.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012983.hg.1 |
ENST00000416175.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012983.hg.1 |
ENST00000416175.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05020695.hg.1 |
TC05001510.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05020695.hg.1 |
ENST00000306388.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05020695.hg.1 |
ENST00000380377.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05020695.hg.1 |
ENST00000517679.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05020695.hg.1 |
ENST00000520361.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05020695.hg.1 |
ENST00000522370.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05020695.hg.1 |
NM_004607.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006914.hg.1 |
ENST00000436520.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006914.hg.1 |
ENST00000436520.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500177800.hg.1 |
ENST00000306388.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500177800.hg.1 |
ENST00000380377.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500177800.hg.1 |
ENST00000518338.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500177800.hg.1 |
ENST00000520039.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500177800.hg.1 |
ENST00000520361.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500177800.hg.1 |
NM_001297738.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500177800.hg.1 |
NM_001297740.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500177800.hg.1 |
NM_004607.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500177800.hg.1 |
ENST00000306388.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500177800.hg.1 |
ENST00000380377.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500177800.hg.1 |
ENST00000518338.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500177800.hg.1 |
ENST00000520039.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500177800.hg.1 |
ENST00000520361.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500177800.hg.1 |
NM_001297738.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500177800.hg.1 |
NM_001297740.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500177800.hg.1 |
NM_004607.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500177800.hg.1 |
TC0500011212.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500177806.hg.1 |
ENST00000380377.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500177806.hg.1 |
NM_001297738.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500177806.hg.1 |
NM_001297740.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500177806.hg.1 |
NM_004607.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500177806.hg.1 |
ENST00000380377.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500177806.hg.1 |
NM_001297738.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500177806.hg.1 |
NM_001297740.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500177806.hg.1 |
NM_004607.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500177806.hg.1 |
TC0500011212.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500011212.hg.1 |
ENST00000306388.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011212.hg.1 |
ENST00000306388.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011212.hg.1 |
ENST00000306388.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011212.hg.1 |
ENST00000306388.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011212.hg.1 |
ENST00000306388.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011212.hg.1 |
ENST00000306388.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011212.hg.1 |
ENST00000306388.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011212.hg.1 |
ENST00000380377.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011212.hg.1 |
ENST00000380377.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011212.hg.1 |
ENST00000380377.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011212.hg.1 |
ENST00000380377.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011212.hg.1 |
ENST00000380377.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011212.hg.1 |
ENST00000380377.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011212.hg.1 |
ENST00000380377.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011212.hg.1 |
ENST00000517679.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011212.hg.1 |
ENST00000517679.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011212.hg.1 |
ENST00000517679.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011212.hg.1 |
ENST00000517679.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011212.hg.1 |
ENST00000517679.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011212.hg.1 |
ENST00000517881.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011212.hg.1 |
ENST00000517881.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011212.hg.1 |
ENST00000517881.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011212.hg.1 |
ENST00000517881.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011212.hg.1 |
ENST00000518218.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011212.hg.1 |
ENST00000518218.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011212.hg.1 |
ENST00000518218.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011212.hg.1 |
ENST00000518218.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011212.hg.1 |
ENST00000518218.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011212.hg.1 |
ENST00000518338.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011212.hg.1 |
ENST00000518338.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011212.hg.1 |
ENST00000518338.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011212.hg.1 |
ENST00000518338.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011212.hg.1 |
ENST00000518338.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011212.hg.1 |
ENST00000518338.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011212.hg.1 |
ENST00000520039.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011212.hg.1 |
ENST00000520039.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011212.hg.1 |
ENST00000520361.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011212.hg.1 |
ENST00000520361.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011212.hg.1 |
ENST00000520361.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011212.hg.1 |
ENST00000520361.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011212.hg.1 |
ENST00000522370.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011212.hg.1 |
ENST00000522370.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011212.hg.1 |
ENST00000522370.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011212.hg.1 |
ENST00000522370.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011212.hg.1 |
NM_001297738.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011212.hg.1 |
NM_001297738.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011212.hg.1 |
NM_001297738.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011212.hg.1 |
NM_001297738.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011212.hg.1 |
NM_001297738.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011212.hg.1 |
NM_001297738.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011212.hg.1 |
NM_001297738.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011212.hg.1 |
NM_001297738.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011212.hg.1 |
NM_001297738.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011212.hg.1 |
NM_001297740.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011212.hg.1 |
NM_001297740.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011212.hg.1 |
NM_001297740.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011212.hg.1 |
NM_001297740.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011212.hg.1 |
NM_001297740.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011212.hg.1 |
NM_001297740.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011212.hg.1 |
NM_004607.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011212.hg.1 |
NM_004607.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011212.hg.1 |
NM_004607.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011212.hg.1 |
NM_004607.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011212.hg.1 |
NM_004607.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011212.hg.1 |
NM_004607.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011212.hg.1 |
NM_004607.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012983.hg.1 |
ENST00000416175.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012983.hg.1 |
ENST00000416175.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500177797.hg.1 |
ENST00000306388.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500177797.hg.1 |
ENST00000380377.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500177797.hg.1 |
ENST00000517679.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500177797.hg.1 |
ENST00000518218.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500177797.hg.1 |
ENST00000518338.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500177797.hg.1 |
ENST00000520361.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500177797.hg.1 |
ENST00000522370.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500177797.hg.1 |
NM_001297738.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500177797.hg.1 |
NM_001297740.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500177797.hg.1 |
NM_004607.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500177797.hg.1 |
ENST00000306388.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500177797.hg.1 |
ENST00000380377.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500177797.hg.1 |
ENST00000517679.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500177797.hg.1 |
ENST00000518218.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500177797.hg.1 |
ENST00000518338.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500177797.hg.1 |
ENST00000520361.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500177797.hg.1 |
ENST00000522370.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500177797.hg.1 |
NM_001297738.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500177797.hg.1 |
NM_001297740.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500177797.hg.1 |
NM_004607.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500177797.hg.1 |
TC0500011212.hg.1 |
Clariom D TC |