Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008907.hg.1 |
ENST00000418945.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008907.hg.1 |
ENST00000418945.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008907.hg.1 |
ENST00000418945.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008907.hg.1 |
ENST00000418945.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008907.hg.1 |
ENST00000418945.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008907.hg.1 |
ENST00000418945.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008907.hg.1 |
ENST00000418945.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008907.hg.1 |
NR_125843.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008907.hg.1 |
NR_125843.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008907.hg.1 |
NR_125843.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008907.hg.1 |
NR_125843.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600161570.hg.1 |
ENST00000418945.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600161570.hg.1 |
NR_125843.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600161570.hg.1 |
ENST00000418945.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600161570.hg.1 |
NR_125843.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600161570.hg.1 |
TC0600008907.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600008907.hg.1 |
ENST00000418945.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008907.hg.1 |
ENST00000418945.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008907.hg.1 |
ENST00000418945.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008907.hg.1 |
ENST00000418945.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008907.hg.1 |
ENST00000418945.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008907.hg.1 |
ENST00000418945.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008907.hg.1 |
ENST00000418945.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008907.hg.1 |
NR_125843.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008907.hg.1 |
NR_125843.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008907.hg.1 |
NR_125843.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008907.hg.1 |
NR_125843.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600161569.hg.1 |
NR_125843.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600161569.hg.1 |
NR_125843.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600161569.hg.1 |
TC0600008907.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600161572.hg.1 |
ENST00000418945.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600161572.hg.1 |
NR_125843.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600161572.hg.1 |
ENST00000418945.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600161572.hg.1 |
NR_125843.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600161572.hg.1 |
TC0600008907.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600161575.hg.1 |
ENST00000418945.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600161575.hg.1 |
NR_125843.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600161575.hg.1 |
ENST00000418945.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600161575.hg.1 |
NR_125843.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600161575.hg.1 |
TC0600008907.hg.1 |
Clariom D TC |