Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007260.hg.1 |
ENST00000610803.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007260.hg.1 |
ENST00000610803.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007260.hg.1 |
ENST00000610803.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007260.hg.1 |
ENST00000610803.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007260.hg.1 |
ENST00000610803.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007260.hg.1 |
ENST00000610803.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007260.hg.1 |
ENST00000610803.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007260.hg.1 |
ENST00000611606.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007260.hg.1 |
ENST00000611606.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007260.hg.1 |
ENST00000611606.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007260.hg.1 |
ENST00000611606.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007260.hg.1 |
ENST00000611606.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007260.hg.1 |
ENST00000611606.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007260.hg.1 |
ENST00000611606.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007260.hg.1 |
ENST00000611606.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007260.hg.1 |
ENST00000611606.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007260.hg.1 |
ENST00000611606.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007260.hg.1 |
ENST00000611606.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007260.hg.1 |
ENST00000615326.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007260.hg.1 |
ENST00000615326.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007260.hg.1 |
ENST00000615326.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007260.hg.1 |
ENST00000615326.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007260.hg.1 |
ENST00000615326.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007260.hg.1 |
ENST00000615326.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007260.hg.1 |
ENST00000615326.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007260.hg.1 |
ENST00000617940.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007260.hg.1 |
ENST00000617940.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007260.hg.1 |
ENST00000617940.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007260.hg.1 |
ENST00000617940.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007260.hg.1 |
ENST00000617940.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007260.hg.1 |
ENST00000617940.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007260.hg.1 |
ENST00000617940.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007260.hg.1 |
ENST00000617940.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007260.hg.1 |
ENST00000618340.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007260.hg.1 |
ENST00000618340.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007260.hg.1 |
ENST00000618340.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007260.hg.1 |
ENST00000618340.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007260.hg.1 |
ENST00000618340.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007260.hg.1 |
ENST00000618340.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007260.hg.1 |
ENST00000618340.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007260.hg.1 |
ENST00000618340.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007260.hg.1 |
ENST00000623489.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007260.hg.1 |
ENST00000623489.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007260.hg.1 |
ENST00000623489.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007260.hg.1 |
ENST00000623489.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007260.hg.1 |
ENST00000624467.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007260.hg.1 |
ENST00000624467.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007260.hg.1 |
NR_034006.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007260.hg.1 |
NR_034006.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007260.hg.1 |
NR_034006.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007260.hg.1 |
NR_034006.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007260.hg.1 |
NR_034006.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007260.hg.1 |
NR_034006.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007260.hg.1 |
NR_034006.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007260.hg.1 |
NR_034006.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007260.hg.1 |
NR_034006.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007260.hg.1 |
NR_034006.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007260.hg.1 |
NR_034006.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007260.hg.1 |
NR_034006.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007260.hg.1 |
ENST00000610803.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007260.hg.1 |
ENST00000610803.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007260.hg.1 |
ENST00000610803.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007260.hg.1 |
ENST00000610803.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007260.hg.1 |
ENST00000610803.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007260.hg.1 |
ENST00000610803.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007260.hg.1 |
ENST00000610803.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007260.hg.1 |
ENST00000611606.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007260.hg.1 |
ENST00000611606.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007260.hg.1 |
ENST00000611606.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007260.hg.1 |
ENST00000611606.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007260.hg.1 |
ENST00000611606.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007260.hg.1 |
ENST00000611606.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007260.hg.1 |
ENST00000611606.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007260.hg.1 |
ENST00000611606.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007260.hg.1 |
ENST00000611606.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007260.hg.1 |
ENST00000611606.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007260.hg.1 |
ENST00000611606.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007260.hg.1 |
ENST00000615326.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007260.hg.1 |
ENST00000615326.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007260.hg.1 |
ENST00000615326.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007260.hg.1 |
ENST00000615326.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007260.hg.1 |
ENST00000615326.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007260.hg.1 |
ENST00000615326.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007260.hg.1 |
ENST00000615326.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007260.hg.1 |
ENST00000617940.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007260.hg.1 |
ENST00000617940.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007260.hg.1 |
ENST00000617940.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007260.hg.1 |
ENST00000617940.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007260.hg.1 |
ENST00000617940.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007260.hg.1 |
ENST00000617940.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007260.hg.1 |
ENST00000617940.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007260.hg.1 |
ENST00000617940.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007260.hg.1 |
ENST00000618340.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007260.hg.1 |
ENST00000618340.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007260.hg.1 |
ENST00000618340.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007260.hg.1 |
ENST00000618340.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007260.hg.1 |
ENST00000618340.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007260.hg.1 |
ENST00000618340.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007260.hg.1 |
ENST00000618340.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007260.hg.1 |
ENST00000618340.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007260.hg.1 |
ENST00000623489.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007260.hg.1 |
ENST00000623489.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007260.hg.1 |
ENST00000623489.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007260.hg.1 |
ENST00000623489.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007260.hg.1 |
ENST00000624467.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007260.hg.1 |
ENST00000624467.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007260.hg.1 |
NR_034006.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007260.hg.1 |
NR_034006.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007260.hg.1 |
NR_034006.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007260.hg.1 |
NR_034006.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007260.hg.1 |
NR_034006.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007260.hg.1 |
NR_034006.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007260.hg.1 |
NR_034006.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007260.hg.1 |
NR_034006.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007260.hg.1 |
NR_034006.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007260.hg.1 |
NR_034006.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007260.hg.1 |
NR_034006.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007260.hg.1 |
NR_034006.2 |
Refseq |