Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDCCAG8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011544021.2 | XP_011542323.1 | 7 - 8 | 827 | 80 | |
XM_011544022.2 | XP_011542324.1 | 7 - 8 | 902 | 80 | ||
XM_011544023.2 | XP_011542325.1 | 7 - 8 | 827 | 80 | ||
XM_011544024.2 | XP_011542326.1 | 7 - 8 | 827 | 80 | ||
XM_011544026.2 | XP_011542328.1 | 7 - 8 | 827 | 80 | ||
XM_011544029.2 | XP_011542331.1 | 7 - 8 | 827 | 80 | ||
XM_017000103.1 | XP_016855592.1 | 7 - 8 | 827 | 80 | ||
XM_017000108.1 | XP_016855597.1 | 7 - 8 | 902 | 80 | ||
GenBank mRNA | AK023586.1 | - | 2 - 3 | 153 | 80 |
Set Membership: |
![]() ![]() ![]() ![]() |