Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001242672.1 | NP_001229601.1 | 5 - 6 | 1031 | 59 | |
XM_016999990.1 | XP_016855479.1 | 5 - 6 | 1103 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Homo sapiens Probe spans exons |