Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30587938 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30587934 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30587937 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30588037 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30587935 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30588036 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30588038 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30588039 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2014978 |
U41285.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2014978 |
NM_007889.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2014978 |
AK052151.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
855969 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
969657 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
256763 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
705606 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
956727 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
736976 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
223449 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
390582 |
AK051895.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
390582 |
AK122329.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
390582 |
BC055691.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
390582 |
BC066146.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
390582 |
BC083181.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
390582 |
NM_172449.2 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
191209 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
131950 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P155764 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P137473 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2014978 |
U41285.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2014978 |
NM_007889.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2014978 |
AK052151.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P480209 |
U41285.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P480209 |
NM_007889.2 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P480209 |
AK052151.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30588036 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30587937 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30587952 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30587939 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30587943 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30587936 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30587945 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30588028 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30587922 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30587946 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30588032 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30587915 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30587940 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30587933 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30588033 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30587948 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30588027 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30587930 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30588030 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30587931 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30587951 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30587935 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30587944 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30588039 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30587926 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30588040 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30588034 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30587918 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30587942 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30587929 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30588038 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30587913 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30587949 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30587947 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P08527201 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P155764 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P08527240 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P137473 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P480209 |
U41285.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P480209 |
NM_007889.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P480209 |
AK052151.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P123134 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P137473 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P155764 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P12674816 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P12674826 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30587935 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30587934 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30588037 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30587937 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30588039 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30587938 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30588038 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30588036 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30588024 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30588056 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30587991 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30588027 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30588092 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30588000 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30588018 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30587944 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30587906 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30587995 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30588020 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30587913 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30587987 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30588006 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30588050 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30588082 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30587897 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30587903 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30588023 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30587911 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30587930 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30588012 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30587951 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30587985 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30588060 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30588105 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30588108 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30587969 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30588033 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30587957 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30588039 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30587988 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30588003 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30588013 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30587962 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30588019 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30588083 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30587936 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30587949 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30587964 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30588036 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30588100 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30588049 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30588069 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30588090 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30588086 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30587907 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30587915 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30588001 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30588052 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30587935 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30587929 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30588022 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30588065 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30587940 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30587948 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30588074 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30587956 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30588034 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30587945 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30587946 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30587989 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30588102 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30587909 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30587994 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30588085 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30588045 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30588095 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30588070 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30588007 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30587905 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30588058 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30588068 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30588028 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30588087 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30588051 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30588078 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30588031 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30588063 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30588041 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30587878 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30588077 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30588075 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30587998 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30587983 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30588055 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30588067 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30588106 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30587939 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30588073 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30587900 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30588115 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30588047 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30588016 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30588080 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30587960 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30587971 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30587899 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30588042 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30587933 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30588040 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30587943 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30588025 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30587966 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30588010 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30588038 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30588030 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30587937 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30588006 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30587991 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30587966 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30588040 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30587952 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12958182 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30587899 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30588024 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30587957 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30588019 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30588038 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30588039 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30587979 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30587971 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12835555 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30587962 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30587927 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12565332 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30588018 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30588030 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30587913 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12674820 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30588031 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12932178 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30588036 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30587909 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30588042 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30587940 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30587933 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30587983 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30588012 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30588000 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30587951 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12793391 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30588022 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30587900 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12820839 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30588041 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30587931 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30587956 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30587954 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12517685 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12696087 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30588016 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30587908 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30587911 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30587903 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30587944 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30588001 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12740447 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30587982 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12674826 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30587989 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30587926 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30587929 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12973116 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30587973 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30588010 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30588023 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30587906 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30588009 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30587960 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30587998 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30587942 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12501335 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30587985 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30587987 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30587943 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30587992 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30587988 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30587930 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30587922 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30587939 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30587925 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30587921 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30587918 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30587947 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30588007 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30587949 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30587948 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30587915 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30588020 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30588034 |
- |
- |