Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29241103 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29241105 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29241102 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29241108 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29241099 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29241101 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29241100 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29241104 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29241109 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29241106 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29241107 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P455932 |
Z48043.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P455932 |
AX785009.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P455932 |
BC025432.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P455932 |
AK158462.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P455932 |
AK160916.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P455932 |
Z48043.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P455932 |
AX785009.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P455932 |
BC025432.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P455932 |
AK158462.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P455932 |
AK160916.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
62861 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
596325 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
752354 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
211294 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
920045 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
12094 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
309617 |
AK045570.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
309617 |
AK162580.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
309617 |
AK163906.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
309617 |
NM_001083806.1 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
309617 |
NM_013540.2 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
59103 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
346326 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
169647 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
98699 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P100869 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P101461 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P455932 |
Z48043.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P455932 |
AX785009.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P455932 |
BC025432.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P455932 |
AK158462.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P455932 |
AK160916.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P455932 |
Z48043.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P455932 |
AX785009.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P455932 |
BC025432.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P455932 |
AK158462.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P455932 |
AK160916.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P455932 |
Z48043.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P455932 |
AX785009.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P455932 |
BC025432.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P455932 |
AK158462.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P455932 |
AK160916.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P455932 |
Z48043.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P455932 |
AX785009.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P455932 |
BC025432.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P455932 |
AK158462.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P455932 |
AK160916.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29241134 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29241133 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29241107 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29241098 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29241125 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29241122 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29241102 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29241112 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29241124 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29241091 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29241104 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29241136 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29241126 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29241116 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29241137 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29241128 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29241110 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29241120 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29241108 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29241131 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29241103 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29241117 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29241115 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29241099 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29241094 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29241101 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29241106 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29241113 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29241096 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P100869 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P101461 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P455932 |
Z48043.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P455932 |
AX785009.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P455932 |
BC025432.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P455932 |
AK158462.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P455932 |
AK160916.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P455932 |
Z48043.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P455932 |
AX785009.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P455932 |
BC025432.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P455932 |
AK158462.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P455932 |
AK160916.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P122909 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P101461 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P100869 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P10865390 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P10628433 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P11122267 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P10628444 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P10628451 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29241105 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29241108 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29241103 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29241102 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29241109 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29241107 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29241106 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29241100 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29241101 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29241104 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29241099 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29240989 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29241065 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29241131 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29241025 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29241045 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29241031 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29241001 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29241091 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29241038 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29241164 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29241104 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29241147 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29241106 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29241063 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29241133 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29241059 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29240991 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29241004 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29241110 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29241029 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29240996 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29241101 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29241027 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29241052 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29241156 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29241146 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29241096 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29241083 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29241076 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29241103 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29241085 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29241068 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29241043 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29241009 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29241162 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29241152 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29241036 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29241039 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29241017 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29241057 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29241098 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29241115 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29241126 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29240993 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29241141 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29241159 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29241122 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29241033 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29241140 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29241013 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29241150 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29241124 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29241007 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29241067 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29241087 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29241128 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29241049 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29241114 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29241117 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29241048 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29241042 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29241144 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29241080 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29241149 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29241123 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29241108 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29241016 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29241003 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29241161 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29241120 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29241158 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29241116 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29241112 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29241113 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29241094 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29241066 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29241056 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29241136 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29241034 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29241117 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10708089 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29241094 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10475424 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29241007 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29241112 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29241003 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29241141 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29241146 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29241102 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29241139 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29241090 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10880381 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29241011 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29241103 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10934183 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29241104 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10720760 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29241087 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29241116 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29241109 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29241126 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29241076 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29241124 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29241131 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29241123 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10708092 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29241101 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29241132 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10873299 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29241147 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29241072 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29241113 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29241110 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29241088 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29241115 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29241096 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29241133 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29241150 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10628436 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29241128 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29241099 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29241114 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29241091 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29241069 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10934182 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29241083 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29241129 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10574826 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29241122 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29241009 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10708090 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10880388 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29241098 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10706510 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29241138 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10873301 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29241120 |
- |
- |