Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phox2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008888.3 | NP_032914.1 | 1 - 2 | 429 | 91 | |
GenBank mRNA | AK052579.2 | - | 1 - 2 | 360 | 91 | |
BC079610.1 | - | 1 - 2 | 429 | 30360139 | 91 | |
Y14493.1 | - | 1 - 2 | 462 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons |