Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00446826_m1
          See other NR5A1 GE Assays ›
          Gene Symbol:
          Nr5a1
          Gene Name:
          nuclear receptor subfamily 5, group A, member 1
          Gene Aliases:
          Ad4BP, ELP, ELP-3, Ftz-F1, Ftzf1, SF-1, SF1
          Chromosome Location:
          Chr.2: 38692656 - 38714560 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00446826_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information
          • Citations (1)

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nr5a1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_139051.3 NP_620639.1 3 - 4 447 77
          GenBank mRNA AB000490.1 - 3 - 4 491 77
          AF511594.1 - 3 - 4 277 77
          AK134378.1 - 3 - 4 448 77
          AK144376.1 - 3 - 4 408 77
          AK156911.1 - 3 - 4 415 77
          BC110476.1 - 3 - 4 390 40047261 77
          BC110477.1 - 3 - 4 390 40047262 77
          D10584.1 - 2 - 3 677 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200003423.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Nuclear Receptor Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nr5a1

          Panther Classification:

          Molecular Function -

          C4 zinc finger nuclear receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          luteinization
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          male gonad development
          tissue development
          multicellular organism aging
          reproductive process
          cell differentiation
          adrenal gland development
          hormone metabolic process
          steroid hormone mediated signaling pathway
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          maintenance of protein location in nucleus
          positive regulation of male gonad development
          negative regulation of female gonad development
          DNA binding
          chromatin binding
          double-stranded DNA binding
          transcription factor activity, sequence-specific DNA binding
          transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
          steroid hormone receptor activity
          protein binding
          phospholipid binding
          zinc ion binding
          lipid binding
          enzyme binding
          sequence-specific DNA binding
          metal ion binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0200029350.mm.1 NM_139051.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200029350.mm.1 NM_139051.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200029350.mm.1 TC0200003423.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200029347.mm.1 NM_139051.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200029347.mm.1 NM_139051.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200029347.mm.1 TC0200003423.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200029348.mm.1 NM_139051.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200029348.mm.1 NM_139051.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200029348.mm.1 TC0200003423.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200029351.mm.1 NM_139051.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200029351.mm.1 NM_139051.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200029351.mm.1 TC0200003423.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 496916 - -

          Back To Top
           

          Citations (1)

          Effects of gestational diethylstilbestrol treatment on male and female gonads during early embryonic development.
          Authors
          Ikeda Y, Tanaka H, Esaki M
          Journal
          Endocrinology - Page No 3970 (Fri Aug 01 00:00:00 EDT 2008)
          Volume (Year)
          149 (2008)
          Product Usage
          -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0