Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il1f9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153511.3 | NP_705731.2 | 3 - 4 | 521 | 68 | |
GenBank mRNA | AK081783.1 | - | 3 - 4 | 516 | 68 | |
AK156687.1 | - | 518 | 68 | |||
AY071843.1 | - | 2 - 3 | 247 | 68 | ||
BC116227.1 | - | 2 - 3 | 247 | 40091521 | 68 | |
BC116228.1 | - | 2 - 3 | 247 | 40091522 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |