Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Per2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011066.3 | NP_035196.2 | 4 - 5 | 612 | 59 | |
GenBank mRNA | AF036893.1 | - | 4 - 5 | 582 | 59 | |
AK044658.1 | - | 4 - 5 | 557 | 59 | ||
AK159847.1 | - | 4 - 5 | 614 | 59 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |