Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arrb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271358.1 | NP_001258287.1 | 2 - 3 | 205 | 62 | |
NM_001271359.1 | NP_001258288.1 | 2 - 3 | 205 | 62 | ||
NM_001271360.1 | NP_001258289.1 | 2 - 3 | 205 | 62 | ||
NM_145429.5 | NP_663404.1 | 2 - 3 | 205 | 62 | ||
GenBank mRNA | AK089542.1 | - | 2 - 3 | 140 | 62 | |
AK150389.1 | - | 2 - 3 | 182 | 62 | ||
AK154874.1 | - | 2 - 3 | 125 | 62 | ||
AK159317.1 | - | 2 - 3 | 133 | 62 | ||
AK162889.1 | - | 2 - 3 | 142 | 62 | ||
AK170889.1 | - | 2 - 3 | 115 | 62 | ||
BC016642.1 | - | 2 - 3 | 87 | 2651372 | 62 | |
BY201367.1 | - | 2 - 3 | 205 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Probe spans exons |