Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adamtsl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001301705.1 | NP_001288634.1 | 14 - 15 | 2744 | 71 | |
NM_144899.3 | NP_659148.2 | 14 - 15 | 2626 | 71 | ||
GenBank mRNA | AK155481.1 | - | 14 - 15 | 2677 | 71 | |
AY158701.1 | - | 14 - 15 | 2567 | 71 | ||
BC016215.1 | - | 1 - 2 | 89 | 71 | ||
BC117925.1 | - | 13 - 14 | 2539 | 40097608 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons |