Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200035430.mm.1 |
NM_175094.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200035430.mm.1 |
NM_175094.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200035430.mm.1 |
TC0200004221.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200035422.mm.1 |
NM_175094.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200035422.mm.1 |
NM_175094.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200035422.mm.1 |
TC0200004221.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200035431.mm.1 |
NM_175094.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200035431.mm.1 |
NM_175094.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200035431.mm.1 |
TC0200004221.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200035434.mm.1 |
NM_175094.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200035434.mm.1 |
NM_175094.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200035434.mm.1 |
TC0200004221.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200035432.mm.1 |
NM_175094.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200035432.mm.1 |
NM_175094.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200035432.mm.1 |
TC0200004221.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200035426.mm.1 |
NM_175094.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200035426.mm.1 |
NM_175094.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200035426.mm.1 |
TC0200004221.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200035427.mm.1 |
NM_175094.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200035427.mm.1 |
NM_175094.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200035427.mm.1 |
TC0200004221.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200035425.mm.1 |
NM_175094.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200035425.mm.1 |
NM_175094.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200035425.mm.1 |
TC0200004221.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200035428.mm.1 |
NM_175094.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200035428.mm.1 |
NM_175094.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200035428.mm.1 |
TC0200004221.mm.1 |
MTA TC |