Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2014882 |
AK089247.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2014882 |
AK080914.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2014882 |
NM_175512.2 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
282245 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
312517 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
540588 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
708101 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
581568 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
358563 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
315119 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
502767 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
464695 |
AK004770.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
464695 |
AK020263.2 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
464695 |
AK080334.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
464695 |
AK143568.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
464695 |
AK169402.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
464695 |
AK172379.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
464695 |
BC029090.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
464695 |
NM_028864.2 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
224567 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
308427 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
224566 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
487632 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
619424 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
174976 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
61104 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P114775 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P150920 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P132444 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2014882 |
AK089247.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2014882 |
AK080914.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2014882 |
NM_175512.2 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P149126 |
BC118029.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P149126 |
AK089247.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P149126 |
AK050180.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P149126 |
AK080914.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P149126 |
NM_175512.2 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21308567 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21308595 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21308558 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21308538 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21308563 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21308566 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21308590 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21308536 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21308569 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21308552 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21308588 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21308579 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21308583 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21308593 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21308543 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21308565 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21308539 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21308553 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21308545 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21308561 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21308535 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21308580 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21308555 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21308574 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21308577 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21308585 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21308556 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21308582 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21308560 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21308594 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21308581 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21308584 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21308541 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21308550 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21308546 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21308572 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P04555769 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P132444 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P114775 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P04555758 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P00396497 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P149126 |
BC118029.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P149126 |
AK089247.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P149126 |
AK050180.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P149126 |
AK080914.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P149126 |
NM_175512.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P572476 |
AK089247.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P106900 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P114775 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P132444 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P01211028 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P01211027 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P01211022 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P01454455 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P01454459 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P01211019 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308740 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308670 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308572 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308583 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308610 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308691 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308737 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308636 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308662 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308548 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308696 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308705 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308590 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308538 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308690 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308735 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308729 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308648 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308556 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308699 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308720 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308678 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308569 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308567 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308655 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308677 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308582 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308614 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308550 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308601 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308607 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308704 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308739 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308577 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308545 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308588 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308652 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308585 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308659 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308560 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308558 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308553 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308661 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308665 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308647 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308573 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308561 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308628 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308683 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308657 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308716 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308694 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308552 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308706 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308753 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308742 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308616 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308689 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308630 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308536 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308638 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308565 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308686 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308593 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308707 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308618 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308693 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308541 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308613 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308580 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308731 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308674 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308680 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308555 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308600 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308605 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308668 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308608 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308637 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308749 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308594 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308712 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308717 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308563 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308651 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308703 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308566 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308746 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308633 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308730 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308650 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308710 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308543 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308604 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308675 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308728 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308626 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308744 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308701 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308595 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308695 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308603 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308644 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308708 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308724 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308646 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308579 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308598 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308624 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21308611 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01216449 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02048699 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21308607 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21308652 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01829331 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21308610 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21308665 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01862065 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21308553 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21308601 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21308584 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01862061 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21308552 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01187232 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21308567 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21308548 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21308572 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01829335 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21308608 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21308598 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21308577 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21308611 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21308580 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21308543 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01211026 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21308595 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21308585 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21308593 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01890037 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21308606 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21308565 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21308538 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21308661 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21308657 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21308536 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01829319 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21308582 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21308557 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21308602 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21308668 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21308566 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21308578 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01890031 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21308573 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21308541 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21308563 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21308534 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21308560 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21308569 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01625069 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21308651 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21308561 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21308604 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21308613 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21308616 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21308655 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01157894 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21308583 |
- |
- |