Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agbl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289656.1 | NP_001276585.1 | 2 - 3 | 748 | 117 | |
NM_001289657.1 | NP_001276586.1 | 2 - 3 | 748 | 117 | ||
NM_001289658.1 | NP_001276587.1 | 2 - 3 | 748 | 117 | ||
NM_178630.4 | NP_848745.2 | 2 - 3 | 748 | 117 | ||
GenBank mRNA | AK018326.1 | - | 3 - 4 | 597 | 117 | |
AK029793.1 | - | 2 - 3 | 749 | 117 | ||
AK029798.1 | - | 2 - 3 | 332 | 117 | ||
AK030378.1 | - | 2 - 3 | 306 | 117 | ||
AK047649.1 | - | 2 - 3 | 305 | 117 | ||
AK054066.1 | - | 1 - 2 | 1779 | 117 | ||
BC028521.1 | - | 2 - 3 | 260 | 117 | ||
BC087897.1 | - | 2 - 3 | 291 | 117 | ||
BC139065.1 | - | 2 - 3 | 200 | 8862087 | 117 | |
DQ867031.1 | - | 2 - 3 | 749 | 117 | ||
DQ867032.1 | - | 2 - 3 | 749 | 117 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |