Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: D630003M21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001131021.2 | NP_001124493.1 | 3 - 4 | 1690 | 74 | |
NM_177657.5 | NP_808325.2 | 3 - 4 | 1690 | 74 | ||
GenBank mRNA | AK052605.1 | - | 3 - 4 | 1689 | 74 | |
AK168367.1 | - | 3 - 4 | 1692 | 74 | ||
BC137959.1 | - | 3 - 4 | 1601 | 8860978 | 74 | |
BC137960.1 | - | 3 - 4 | 1601 | 8860979 | 74 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |