Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500016497.mm.1 |
TC0500001779.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500016497.mm.1 |
NM_153599.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500016497.mm.1 |
NM_153599.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500016501.mm.1 |
TC0500001779.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500016501.mm.1 |
NM_153599.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500016501.mm.1 |
NM_153599.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500016495.mm.1 |
TC0500001779.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500016495.mm.1 |
NM_153599.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500016495.mm.1 |
NM_153599.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500016498.mm.1 |
TC0500001779.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500016498.mm.1 |
NM_153599.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500016498.mm.1 |
NM_153599.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500016502.mm.1 |
TC0500001779.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500016502.mm.1 |
NM_153599.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500016502.mm.1 |
NM_153599.3 |
Refseq |