Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpep2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001301204.1 | NP_001288133.1 | 5 - 6 | 845 | 54 | |
NM_001301205.1 | NP_001288134.1 | 4 - 5 | 530 | 54 | ||
NM_176913.4 | NP_795887.2 | 5 - 6 | 969 | 54 | ||
GenBank mRNA | AF488552.1 | - | 5 - 6 | 969 | 54 | |
AK089165.1 | - | 2 - 3 | 2583 | 54 | ||
AK089244.1 | - | 5 - 6 | 831 | 54 | ||
AK155099.1 | - | 5 - 6 | 797 | 54 | ||
AK155308.1 | - | 5 - 6 | 796 | 54 | ||
AK155417.1 | - | 5 - 6 | 800 | 54 | ||
AK170094.1 | - | 5 - 6 | 797 | 54 | ||
AK170970.1 | - | 5 - 6 | 797 | 54 | ||
AK171044.1 | - | 5 - 6 | 797 | 54 | ||
AK171122.1 | - | 5 - 6 | 797 | 54 | ||
AK171155.1 | - | 4 - 5 | 510 | 54 | ||
AK171196.1 | - | 5 - 6 | 797 | 54 | ||
AK171268.1 | - | 5 - 6 | 797 | 54 | ||
AK171503.1 | - | 5 - 6 | 797 | 54 | ||
AK171606.1 | - | 5 - 6 | 813 | 54 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Detect Genomic DNA Mus musculus Probe spans exons |