Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Gene Expression
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Gene Expression
  • Microbe & ABR Detection
  • Copy Number Variation
  • Mutation Detection
  • miRNA
  • SNP Genotyping
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central
            This product has been added to your favorites list. Go to My Favorites

            System Message

            OKCancel
            LOADING ...
            • Home
            • › Search Tool
            • › Search Results
            • › Mm00836158_s1
            See other VMN1R18 GE Assays ›
            Gene Symbol:
            Vmn1r18
            Gene Name:
            vomeronasal 1 receptor 18
            Gene Aliases:
            V1rc26
            Chromosome Location:
            Chr.6: 57389668 - 57390567 on Build GRCm38
            Species:
            Mouse
            Species Specific ID (Flybase ID):
            -
            Assay ID Mm00836158_s1
            Best Coverage
            Size
            Availability Inventoried
            Catalog # 4331182
            Price (USD) 272.00
            Your Price
            Online offer (USD):256.65
            256.65
            Check your price ›
            • Genomic Map
            • Assay Details
            • More Information

            Genomic Map

            LOADING... Fetching data..
            ×
            Back To Top

            Assay Details

            Gene Symbol: Vmn1r18
            Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
            RefSeq NM_134181.1 NP_598942.1 1 750 162
            GenBank mRNA BC132061.1 - 1 849 40130338 162
            BC138409.1 - 1 849 8861429 162

            Back To Top

            More Information


            Additional Information:

            Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

            Set Membership:

            Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

            Matching Ambion siRNAs:

            Click here to find siRNAs for Gene Vmn1r18

            Panther Classification:

            Molecular Function -

            transmembrane signal receptor

            Gene Ontology Categories:

            Function(s) Process(es)

            G-protein coupled receptor signaling pathway
            response to pheromone
            pheromone binding
            pheromone receptor activity

            MicroArray Probes:

            Agilent Affymetrix ABI Illumina

            Array Name Probe IDs Targets Target Type
            Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2024894 BC132061.1 mouse_transcriptome
            Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2024894 BC138409.1 mouse_transcriptome
            Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2024894 NM_134181.1 mouse_transcriptome
            Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1980174 BC132061.1 mouse_transcriptome
            Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1980174 BC138409.1 mouse_transcriptome
            Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1980174 NM_134181.1 mouse_transcriptome
            SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1980174 BC132061.1 mouse_transcriptome
            SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1980174 BC138409.1 mouse_transcriptome
            SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1980174 NM_134181.1 mouse_transcriptome
            SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2024894 BC132061.1 mouse_transcriptome
            SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2024894 BC138409.1 mouse_transcriptome
            SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2024894 NM_134181.1 mouse_transcriptome
            Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P452936 BC132061.1 mouse_transcriptome
            Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P452936 BC138409.1 mouse_transcriptome
            Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P452936 NM_134181.1 mouse_transcriptome
            Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24478631 - -
            Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24478636 - -
            Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24478673 - -
            Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24478614 - -
            Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24478616 - -
            Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24478644 - -
            Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24478639 - -
            Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24478676 - -
            Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24478608 - -
            Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24478633 - -
            Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24478628 - -
            Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24478660 - -
            Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24478630 - -
            Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24478635 - -
            Whole Mouse Genome G4122A A_51_P452936 BC132061.1 mouse_transcriptome
            Whole Mouse Genome G4122A A_51_P452936 BC138409.1 mouse_transcriptome
            Whole Mouse Genome G4122A A_51_P452936 NM_134181.1 mouse_transcriptome
            Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P137281 - -
            Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05630718 - -
            Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05630716 - -
            Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05630707 - -
            Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05630699 - -
            Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24478633 - -
            Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24478676 - -
            Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24478631 - -
            Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24478616 - -
            Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24478635 - -
            Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24478630 - -
            Agilent Six_ChIP_02_of_03_v2 A_68_P05630710 - -
            Agilent Six_ChIP_02_of_03_v2 A_68_P24478660 - -
            Agilent Six_ChIP_02_of_03_v2 A_68_P05630707 - -
            Agilent Six_ChIP_02_of_03_v2 A_68_P05630716 - -
            Agilent Six_ChIP_02_of_03_v2 A_68_P24478630 - -
            Agilent Six_ChIP_02_of_03_v2 A_68_P24478666 - -
            Agilent Six_ChIP_02_of_03_v2 A_68_P24478670 - -
            Agilent Six_ChIP_02_of_03_v2 A_68_P05324555 - -
            Agilent Six_ChIP_02_of_03_v2 A_68_P24478644 - -
            Agilent Six_ChIP_02_of_03_v2 A_68_P24478614 - -
            Agilent Six_ChIP_02_of_03_v2 A_68_P24478655 - -
            Agilent Six_ChIP_02_of_03_v2 A_68_P24478607 - -
            Agilent Six_ChIP_02_of_03_v2 A_68_P24478636 - -
            Agilent Six_ChIP_02_of_03_v2 A_68_P24478628 - -
            Agilent Six_ChIP_02_of_03_v2 A_68_P05630709 - -
            Agilent Six_ChIP_02_of_03_v2 A_68_P24478616 - -
            Agilent Six_ChIP_02_of_03_v2 A_68_P24478639 - -
            Agilent Six_ChIP_02_of_03_v2 A_68_P05630718 - -
            Agilent Six_ChIP_02_of_03_v2 A_68_P24478635 - -
            Agilent Six_ChIP_02_of_03_v2 A_68_P24478611 - -
            Agilent Six_ChIP_02_of_03_v2 A_68_P24478633 - -
            Array Name Probe IDs Targets Target Type
            Affymetrix Mouse Gene 1.0 ST Array 10545000 - -
            Affymetrix Mouse Gene 1.1 ST Array 10544999 - -
            Affymetrix Mouse Clariom S Array PSR0600018324.mm.1 TC0600002348.mm.1 MTA TC
            Affymetrix Mouse Clariom S Array PSR0600018324.mm.1 NM_134181.1.1 Refseq
            Affymetrix Mouse Clariom S Array PSR0600018324.mm.1 NM_134181.1 Refseq
            Affymetrix Mouse Clariom S Array PSR0600018325.mm.1 TC0600002348.mm.1 MTA TC
            Affymetrix Mouse Clariom S Array PSR0600018323.mm.1 TC0600002348.mm.1 MTA TC
            Array Name Probe IDs Targets Target Type
            ABI Mouse Genome Survey Microarray 624959 - -
            Array Name Probe IDs Targets Target Type
            Illumina mouse-6 v1.0 expression beadchip 2320164 - -
            Illumina mouseRef-8 v1.0 expression beadchip 0002320164 - -
            Illumina mouseRef-8 v1.1 expression beadchip ILMN_1240473 - -
            Illumina mouseRef-8 v1.1 expression beadchip ILMN_2456634 - -
            Illumina mouse-6 v1.1 expression beadchip ILMN_1240473 - -
            Illumina mouse-6 v1.1 expression beadchip ILMN_2456634 - -
            mouse-6 v1.1 (Illumina) scl00171199.1_296-S - -
            Illumina Sentrix mouse-6.v1 bead array 2320164 - -
            Illumina Sentrix MouseRef-8 Expression BeadChip scl00171199.1_296-S - -
            Illumina Sentrix Mouse-6 Expression BeadChip (2) 2320164 - -
            MouseRef-8 v2 Expression BeadChip ILMN_1240473 - -
            Illumina MouseWG-6 v2.0 expression beadchip ILMN_1240473 - -

            Back To Top

            Related Products

            • TaqMan® Fast Advanced Master Mix
            • SuperScript® VILO™ cDNA Synthesis Kit
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            united states flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-6ff95d844f-tw7ck:80/100.66.77.154:80.
            git-commit: f44aa9153aef922f8fad9a05e87b8aee811da128
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.28.0-Offline