Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Naalad2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028279.3 | NP_082555.2 | 5 - 6 | 88 | ||
GenBank mRNA | AK133611.1 | - | 5 - 6 | 88 | ||
AK135653.1 | - | 7 - 8 | 88 | |||
AK161457.1 | - | 8 - 9 | 971 | 88 | ||
AY243507.1 | - | 5 - 6 | 88 | |||
BC108995.1 | - | 5 - 6 | 40047859 | 88 | ||
BC108996.1 | - | 5 - 6 | 40047860 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |