Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps26b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178027.4 | NP_821170.1 | 4 - 5 | 1192 | 93 | |
GenBank mRNA | AK010160.1 | - | 4 - 5 | 1126 | 93 | |
AK031589.1 | - | 4 - 5 | 1195 | 93 | ||
AK035321.1 | - | 4 - 5 | 1143 | 93 | ||
AK150929.1 | - | 4 - 5 | 1113 | 93 | ||
AK155578.1 | - | 4 - 5 | 1153 | 93 | ||
AK171154.1 | - | 4 - 5 | 1151 | 93 | ||
AK215961.1 | - | 2 - 3 | 237 | 93 | ||
BC029758.1 | - | 2 - 3 | 285 | 93 | ||
BC049180.1 | - | 4 - 5 | 1155 | 6314347 | 93 | |
BC052721.1 | - | 4 - 5 | 1116 | 6809853 | 93 | |
BC062972.1 | - | 4 - 5 | 1116 | 6809878 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |