Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039048.2 | NP_001034137.2 | 2 - 3 | 441 | 73 | |
GenBank mRNA | BC147663.1 | - | 2 - 3 | 409 | 9087452 | 73 |
BC147666.1 | - | 2 - 3 | 409 | 9087455 | 73 | |
CJ131783.1 | - | 2 - 3 | 443 | 73 | ||
CJ131880.1 | - | 2 - 3 | 442 | 73 | ||
DQ229108.1 | - | 2 - 3 | 337 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |